Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7877895-7878168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTRR
Entrez Gene ID
4552
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase reductase
Gene Aliases
MSR, cblE
UniGene
Hs.481551
Gene Chromosome Location
Chr.5: 7869104-7901124 on Build GRCh38
Transcripts
NM_002454.2 NM_024010.2 NR_134480.1 NR_134481.1 NR_134482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5424001 c.554G>C p.R185T Chr.5: 7878015-7878015 on Build GRCh38
1695367 c.583C>T p.P195S Chr.5: 7878044-7878044 on Build GRCh38
6561246 c.483-6G>T p.? Chr.5: 7877938-7877938 on Build GRCh38
3669476 c.568A>G p.I190V Chr.5: 7878029-7878029 on Build GRCh38
7226948 c.573T>A p.S191R Chr.5: 7878034-7878034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747535153 Chr.5: 7878126-7878126 on Build GRCh38
rs908506872 Chr.5: 7877990-7877990 on Build GRCh38
rs780451901 Chr.5: 7878124-7878124 on Build GRCh38
rs147742177 Chr.5: 7878047-7878047 on Build GRCh38
rs368654030 Chr.5: 7877946-7877946 on Build GRCh38
rs996966324 Chr.5: 7878084-7878084 on Build GRCh38
rs769172296 Chr.5: 7878127-7878127 on Build GRCh38
rs748186027 Chr.5: 7878131-7878131 on Build GRCh38
rs114253881 Chr.5: 7878038-7878038 on Build GRCh38
rs777364593 Chr.5: 7878115-7878115 on Build GRCh38
rs779803224 Chr.5: 7878032-7878032 on Build GRCh38
rs141053952 Chr.5: 7878050-7878050 on Build GRCh38
rs143978300 Chr.5: 7878080-7878080 on Build GRCh38
rs553846662 Chr.5: 7877964-7877964 on Build GRCh38
rs747865209 Chr.5: 7878040-7878040 on Build GRCh38
rs774692169 Chr.5: 7878117-7878117 on Build GRCh38
rs161870 Chr.5: 7878079-7878079 on Build GRCh38
rs773514527 Chr.5: 7878136-7878136 on Build GRCh38
rs565590605 Chr.5: 7877924-7877924 on Build GRCh38
rs937816616 Chr.5: 7878022-7878022 on Build GRCh38
rs771216289 Chr.5: 7878036-7878037 on Build GRCh38
rs773160237 Chr.5: 7878071-7878071 on Build GRCh38
rs142714881 Chr.5: 7877988-7877988 on Build GRCh38
rs867974337 Chr.5: 7877983-7877983 on Build GRCh38
rs192942300 Chr.5: 7877925-7877925 on Build GRCh38
rs763322201 Chr.5: 7878023-7878025 on Build GRCh38
rs776844185 Chr.5: 7878061-7878061 on Build GRCh38
rs769406733 Chr.5: 7878041-7878041 on Build GRCh38
rs746183496 Chr.5: 7878046-7878046 on Build GRCh38
rs1532268 Chr.5: 7878066-7878066 on Build GRCh38
rs139333376 Chr.5: 7878037-7878037 on Build GRCh38
rs752182014 Chr.5: 7878101-7878101 on Build GRCh38
rs762774379 Chr.5: 7878077-7878077 on Build GRCh38
rs149037732 Chr.5: 7878082-7878082 on Build GRCh38
rs367609729 Chr.5: 7878049-7878049 on Build GRCh38
rs886060804 Chr.5: 7878029-7878029 on Build GRCh38
rs750209286 Chr.5: 7877997-7877997 on Build GRCh38
rs754157293 Chr.5: 7877947-7877947 on Build GRCh38
rs748533060 Chr.5: 7878138-7878138 on Build GRCh38
rs919772858 Chr.5: 7878009-7878009 on Build GRCh38
rs765530344 Chr.5: 7878067-7878067 on Build GRCh38
rs116439533 Chr.5: 7877974-7877974 on Build GRCh38
rs556611332 Chr.5: 7878005-7878005 on Build GRCh38
rs757635349 Chr.5: 7878045-7878045 on Build GRCh38
rs766155854 Chr.5: 7878143-7878143 on Build GRCh38
rs143058455 Chr.5: 7878116-7878116 on Build GRCh38
rs111414636 Chr.5: 7877989-7877989 on Build GRCh38
rs747099858 Chr.5: 7878125-7878125 on Build GRCh38
rs758631703 Chr.5: 7878118-7878118 on Build GRCh38
rs776897289 Chr.5: 7878130-7878130 on Build GRCh38
rs150853519 Chr.5: 7878015-7878015 on Build GRCh38
rs753947379 Chr.5: 7877965-7877965 on Build GRCh38
rs755787285 Chr.5: 7878112-7878112 on Build GRCh38
rs973962567 Chr.5: 7878008-7878008 on Build GRCh38

View Full Product Details