Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50719758-50720029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LUC7L3
Entrez Gene ID
51747
Gene Name
LUC7 like 3 pre-mRNA splicing factor
Gene Aliases
CRA, CREAP-1, CROP, LUC7A, OA48-18, hLuc7A
UniGene
Hs.130293
Gene Chromosome Location
Chr.17: 50719565-50754269 on Build GRCh38
Transcripts
NM_006107.3 NM_016424.4 XM_005257448.1 XM_005257449.2 XM_005257452.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4067768 c.91C>A p.H31N Chr.17: 50719823-50719823 on Build GRCh38
6705159 c.33A>G p.L11L Chr.17: 50719765-50719765 on Build GRCh38
4067769 c.91C>A p.H31N Chr.17: 50719823-50719823 on Build GRCh38
3983547 c.94G>C p.E32Q Chr.17: 50719826-50719826 on Build GRCh38
3983546 c.94G>C p.E32Q Chr.17: 50719826-50719826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762156104 Chr.17: 50719838-50719838 on Build GRCh38
rs750844101 Chr.17: 50719844-50719844 on Build GRCh38
rs889663306 Chr.17: 50719917-50719917 on Build GRCh38
rs777397233 Chr.17: 50719866-50719866 on Build GRCh38
rs565603102 Chr.17: 50719914-50719914 on Build GRCh38
rs375101535 Chr.17: 50719929-50719929 on Build GRCh38
rs764272372 Chr.17: 50719823-50719823 on Build GRCh38
rs753861011 Chr.17: 50719825-50719825 on Build GRCh38
rs758793799 Chr.17: 50719847-50719847 on Build GRCh38
rs972826739 Chr.17: 50719934-50719934 on Build GRCh38
rs953081469 Chr.17: 50719959-50719959 on Build GRCh38
rs1054582764 Chr.17: 50719839-50719839 on Build GRCh38
rs113260284 Chr.17: 50719852-50719852 on Build GRCh38
rs1046502013 Chr.17: 50719882-50719882 on Build GRCh38
rs375563053 Chr.17: 50719816-50719816 on Build GRCh38
rs949555593 Chr.17: 50719868-50719868 on Build GRCh38
rs761365008 Chr.17: 50719911-50719911 on Build GRCh38
rs534611119 Chr.17: 50719915-50719915 on Build GRCh38
rs916825268 Chr.17: 50719988-50719988 on Build GRCh38
rs745464670 Chr.17: 50719880-50719880 on Build GRCh38
rs991073585 Chr.17: 50719960-50719960 on Build GRCh38
rs752075553 Chr.17: 50719857-50719857 on Build GRCh38
rs1008056498 Chr.17: 50719923-50719923 on Build GRCh38
rs765358967 Chr.17: 50719841-50719841 on Build GRCh38
rs1019402795 Chr.17: 50719925-50719925 on Build GRCh38
rs780348318 Chr.17: 50719849-50719849 on Build GRCh38
rs916710740 Chr.17: 50719845-50719845 on Build GRCh38
rs893056918 Chr.17: 50719908-50719908 on Build GRCh38
rs748723087 Chr.17: 50719870-50719870 on Build GRCh38
rs751633695 Chr.17: 50719845-50719846 on Build GRCh38
rs554699644 Chr.17: 50719975-50719975 on Build GRCh38
rs756973442 Chr.17: 50719874-50719874 on Build GRCh38
rs773902055 Chr.17: 50719873-50719873 on Build GRCh38
rs551776463 Chr.17: 50719891-50719891 on Build GRCh38
rs369589389 Chr.17: 50719939-50719939 on Build GRCh38
rs755412112 Chr.17: 50719865-50719865 on Build GRCh38
rs397713660 Chr.17: 50719851-50719852 on Build GRCh38
rs11398903 Chr.17: 50719849-50719850 on Build GRCh38
rs1028162834 Chr.17: 50719916-50719916 on Build GRCh38
rs898202166 Chr.17: 50719956-50719956 on Build GRCh38
rs778461001 Chr.17: 50719877-50719877 on Build GRCh38
rs371275354 Chr.17: 50719787-50719787 on Build GRCh38
rs949585144 Chr.17: 50719995-50719995 on Build GRCh38
rs902594486 Chr.17: 50719889-50719889 on Build GRCh38
rs1053770168 Chr.17: 50719899-50719899 on Build GRCh38

View Full Product Details