Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153297927-153298174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGLYRP3
Entrez Gene ID
114771
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
PGLYRPIalpha, PGRP-Ialpha, PGRPIA
UniGene
Hs.348266
Gene Chromosome Location
Chr.1: 153297589-153312975 on Build GRCh38
Transcripts
NM_052891.2 XM_011509118.1 XM_011509119.1 XM_011509120.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983864 c.999C>G p.I333M Chr.1: 153297983-153297983 on Build GRCh38
6121141 c.886G>T p.D296Y Chr.1: 153298096-153298096 on Build GRCh38
2207598 c.869C>T p.A290V Chr.1: 153298113-153298113 on Build GRCh38
6058079 c.969G>A p.L323L Chr.1: 153298013-153298013 on Build GRCh38
2207597 c.870G>A p.A290A Chr.1: 153298112-153298112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185029783 Chr.1: 153298078-153298078 on Build GRCh38
rs374263961 Chr.1: 153298037-153298037 on Build GRCh38
rs200006086 Chr.1: 153298151-153298151 on Build GRCh38
rs190366105 Chr.1: 153298138-153298138 on Build GRCh38
rs758678036 Chr.1: 153297954-153297954 on Build GRCh38
rs374418121 Chr.1: 153298143-153298143 on Build GRCh38
rs778767559 Chr.1: 153298092-153298092 on Build GRCh38
rs763363784 Chr.1: 153298147-153298147 on Build GRCh38
rs146741860 Chr.1: 153298069-153298069 on Build GRCh38
rs780880043 Chr.1: 153298134-153298134 on Build GRCh38
rs762385160 Chr.1: 153298047-153298047 on Build GRCh38
rs751839832 Chr.1: 153298070-153298070 on Build GRCh38
rs775325906 Chr.1: 153298022-153298022 on Build GRCh38
rs552648838 Chr.1: 153298098-153298098 on Build GRCh38
rs200719839 Chr.1: 153298059-153298059 on Build GRCh38
rs374030758 Chr.1: 153298112-153298112 on Build GRCh38
rs544597913 Chr.1: 153298079-153298079 on Build GRCh38
rs779837106 Chr.1: 153298107-153298107 on Build GRCh38
rs756935845 Chr.1: 153298010-153298010 on Build GRCh38
rs138438992 Chr.1: 153297985-153297985 on Build GRCh38
rs369502719 Chr.1: 153298011-153298011 on Build GRCh38
rs371894209 Chr.1: 153298140-153298140 on Build GRCh38
rs766584893 Chr.1: 153298067-153298067 on Build GRCh38
rs994367395 Chr.1: 153298066-153298066 on Build GRCh38
rs745466234 Chr.1: 153298014-153298014 on Build GRCh38
rs542269185 Chr.1: 153298003-153298003 on Build GRCh38
rs757245633 Chr.1: 153298091-153298091 on Build GRCh38
rs768337483 Chr.1: 153298028-153298028 on Build GRCh38
rs750203950 Chr.1: 153298103-153298103 on Build GRCh38
rs771736264 Chr.1: 153298020-153298020 on Build GRCh38
rs199849093 Chr.1: 153298113-153298113 on Build GRCh38
rs779963621 Chr.1: 153298111-153298111 on Build GRCh38
rs773646455 Chr.1: 153298064-153298064 on Build GRCh38
rs753739167 Chr.1: 153298083-153298083 on Build GRCh38
rs746509898 Chr.1: 153298025-153298025 on Build GRCh38
rs142436988 Chr.1: 153298104-153298104 on Build GRCh38
rs750543110 Chr.1: 153297949-153297949 on Build GRCh38
rs777676470 Chr.1: 153297986-153297986 on Build GRCh38
rs766319038 Chr.1: 153297971-153297971 on Build GRCh38
rs745672504 Chr.1: 153298043-153298043 on Build GRCh38
rs749236657 Chr.1: 153297990-153297990 on Build GRCh38
rs751615092 Chr.1: 153297983-153297983 on Build GRCh38
rs1043074601 Chr.1: 153297965-153297965 on Build GRCh38
rs954911183 Chr.1: 153298121-153298121 on Build GRCh38

View Full Product Details