Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158707157-158707382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYTL3
Entrez Gene ID
94120
Gene Name
synaptotagmin like 3
Gene Aliases
SLP3
UniGene
Hs.436977
Gene Chromosome Location
Chr.6: 158644881-158766298 on Build GRCh38
Transcripts
NM_001009991.3 NM_001242384.1 NM_001242394.1 NM_001242395.1 NM_001318745.1 XM_005267215.4 XM_005267218.4 XM_006715605.3 XM_006715606.3 XM_006715611.3 XM_011536254.2 XM_017011495.1 XM_017011496.1 XM_017011497.1 XM_017011498.1 XM_017011499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889458 c.403G>T p.E135* Chr.6: 158707238-158707238 on Build GRCh38
6406073 c.436C>A p.Q146K Chr.6: 158707271-158707271 on Build GRCh38
6406072 c.436C>A p.Q146K Chr.6: 158707271-158707271 on Build GRCh38
4889459 c.403G>T p.E135* Chr.6: 158707238-158707238 on Build GRCh38
4156020 c.395-7C>G p.? Chr.6: 158707223-158707223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916658119 Chr.6: 158707335-158707335 on Build GRCh38
rs373009593 Chr.6: 158707326-158707326 on Build GRCh38
rs748853493 Chr.6: 158707271-158707271 on Build GRCh38
rs372216226 Chr.6: 158707213-158707213 on Build GRCh38
rs752084471 Chr.6: 158707305-158707305 on Build GRCh38
rs750153707 Chr.6: 158707207-158707207 on Build GRCh38
rs761607259 Chr.6: 158707205-158707205 on Build GRCh38
rs778423151 Chr.6: 158707230-158707230 on Build GRCh38
rs752650712 Chr.6: 158707303-158707303 on Build GRCh38
rs901593929 Chr.6: 158707347-158707347 on Build GRCh38
rs771403624 Chr.6: 158707262-158707262 on Build GRCh38
rs778814744 Chr.6: 158707223-158707223 on Build GRCh38
rs370111316 Chr.6: 158707306-158707306 on Build GRCh38
rs748983706 Chr.6: 158707318-158707318 on Build GRCh38
rs747439589 Chr.6: 158707235-158707235 on Build GRCh38
rs759624935 Chr.6: 158707296-158707296 on Build GRCh38
rs778625081 Chr.6: 158707328-158707328 on Build GRCh38
rs761745333 Chr.6: 158707280-158707280 on Build GRCh38
rs768128134 Chr.6: 158707319-158707319 on Build GRCh38
rs747225962 Chr.6: 158707260-158707261 on Build GRCh38
rs551332776 Chr.6: 158707225-158707225 on Build GRCh38
rs199854012 Chr.6: 158707215-158707215 on Build GRCh38
rs372154726 Chr.6: 158707202-158707202 on Build GRCh38
rs574189044 Chr.6: 158707226-158707226 on Build GRCh38
rs544778801 Chr.6: 158707355-158707355 on Build GRCh38
rs548805403 Chr.6: 158707276-158707276 on Build GRCh38
rs760512434 Chr.6: 158707288-158707288 on Build GRCh38
rs374708271 Chr.6: 158707206-158707206 on Build GRCh38
rs768236018 Chr.6: 158707272-158707272 on Build GRCh38
rs773119026 Chr.6: 158707285-158707285 on Build GRCh38
rs758541648 Chr.6: 158707304-158707304 on Build GRCh38
rs772062348 Chr.6: 158707281-158707281 on Build GRCh38
rs773212044 Chr.6: 158707253-158707253 on Build GRCh38
rs748197982 Chr.6: 158707195-158707200 on Build GRCh38
rs765369589 Chr.6: 158707301-158707301 on Build GRCh38
rs763484697 Chr.6: 158707193-158707193 on Build GRCh38
rs181294151 Chr.6: 158707270-158707270 on Build GRCh38
rs773389158 Chr.6: 158707306-158707307 on Build GRCh38
rs747712287 Chr.6: 158707330-158707330 on Build GRCh38
rs766057389 Chr.6: 158707292-158707292 on Build GRCh38
rs367788987 Chr.6: 158707208-158707208 on Build GRCh38
rs772299001 Chr.6: 158707212-158707213 on Build GRCh38
rs781362173 Chr.6: 158707317-158707317 on Build GRCh38

View Full Product Details