Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132225278-132225539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK32C
Entrez Gene ID
282974
Gene Name
serine/threonine kinase 32C
Gene Aliases
PKE, YANK3
UniGene
Hs.469002
Gene Chromosome Location
Chr.10: 132207482-132332559 on Build GRCh38
Transcripts
NM_001318878.1 NM_001318879.1 NM_173575.3 NR_134911.1 XM_011539688.1 XM_011539690.2 XM_011539693.2 XM_011539694.1 XM_011539695.1 XM_011539696.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6792033 c.819C>A p.S273S Chr.10: 132225290-132225290 on Build GRCh38
6792018 c.811+8C>T p.? Chr.10: 132225519-132225519 on Build GRCh38
5703510 c.812-2A>G p.? Chr.10: 132225338-132225338 on Build GRCh38
6792009 c.815C>T p.P272L Chr.10: 132225333-132225333 on Build GRCh38
5703511 c.773-2A>G p.? Chr.10: 132225338-132225338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765710638 Chr.10: 132225381-132225381 on Build GRCh38
rs775819142 Chr.10: 132225501-132225501 on Build GRCh38
rs183535649 Chr.10: 132225404-132225404 on Build GRCh38
rs550012378 Chr.10: 132225365-132225365 on Build GRCh38
rs756174619 Chr.10: 132225403-132225403 on Build GRCh38
rs767283408 Chr.10: 132225397-132225397 on Build GRCh38
rs976023307 Chr.10: 132225389-132225389 on Build GRCh38
rs951939255 Chr.10: 132225375-132225375 on Build GRCh38
rs925064648 Chr.10: 132225429-132225429 on Build GRCh38
rs752035680 Chr.10: 132225349-132225351 on Build GRCh38
rs771151910 Chr.10: 132225416-132225416 on Build GRCh38
rs41309399 Chr.10: 132225384-132225384 on Build GRCh38
rs781351337 Chr.10: 132225488-132225488 on Build GRCh38
rs201790118 Chr.10: 132225519-132225519 on Build GRCh38
rs766430006 Chr.10: 132225469-132225469 on Build GRCh38
rs773908161 Chr.10: 132225447-132225447 on Build GRCh38
rs760386922 Chr.10: 132225517-132225517 on Build GRCh38
rs772935821 Chr.10: 132225516-132225516 on Build GRCh38
rs771886088 Chr.10: 132225515-132225515 on Build GRCh38
rs377207517 Chr.10: 132225390-132225390 on Build GRCh38
rs753880048 Chr.10: 132225471-132225471 on Build GRCh38
rs191613482 Chr.10: 132225371-132225371 on Build GRCh38
rs761340807 Chr.10: 132225510-132225510 on Build GRCh38
rs576933822 Chr.10: 132225425-132225425 on Build GRCh38
rs1025235506 Chr.10: 132225363-132225363 on Build GRCh38
rs777752577 Chr.10: 132225474-132225474 on Build GRCh38
rs769853252 Chr.10: 132225351-132225351 on Build GRCh38
rs762196423 Chr.10: 132225297-132225297 on Build GRCh38
rs756922162 Chr.10: 132225395-132225395 on Build GRCh38
rs752171583 Chr.10: 132225318-132225318 on Build GRCh38
rs1013884829 Chr.10: 132225348-132225348 on Build GRCh38
rs781202657 Chr.10: 132225342-132225342 on Build GRCh38
rs142198938 Chr.10: 132225332-132225332 on Build GRCh38
rs375197502 Chr.10: 132225370-132225370 on Build GRCh38
rs202230396 Chr.10: 132225521-132225521 on Build GRCh38
rs779177614 Chr.10: 132225359-132225359 on Build GRCh38
rs747257506 Chr.10: 132225415-132225415 on Build GRCh38
rs767260088 Chr.10: 132225464-132225464 on Build GRCh38
rs750215616 Chr.10: 132225400-132225400 on Build GRCh38
rs769176245 Chr.10: 132225503-132225503 on Build GRCh38
rs770087661 Chr.10: 132225496-132225496 on Build GRCh38
rs762540233 Chr.10: 132225430-132225430 on Build GRCh38
rs148669059 Chr.10: 132225313-132225313 on Build GRCh38
rs751622241 Chr.10: 132225520-132225520 on Build GRCh38
rs746234897 Chr.10: 132225493-132225493 on Build GRCh38
rs772848707 Chr.10: 132225500-132225500 on Build GRCh38
rs887876212 Chr.10: 132225326-132225326 on Build GRCh38
rs545953249 Chr.10: 132225433-132225433 on Build GRCh38
rs201852787 Chr.10: 132225309-132225309 on Build GRCh38
rs777891156 Chr.10: 132225413-132225413 on Build GRCh38
rs754936936 Chr.10: 132225473-132225473 on Build GRCh38
rs373281127 Chr.10: 132225518-132225518 on Build GRCh38
rs760019912 Chr.10: 132225311-132225311 on Build GRCh38
rs12411723 Chr.10: 132225308-132225308 on Build GRCh38
rs755936539 Chr.10: 132225468-132225468 on Build GRCh38
rs762871806 Chr.10: 132225506-132225506 on Build GRCh38
rs200927472 Chr.10: 132225299-132225299 on Build GRCh38
rs151066427 Chr.10: 132225343-132225343 on Build GRCh38
rs760905509 Chr.10: 132225305-132225305 on Build GRCh38
rs554645198 Chr.10: 132225383-132225383 on Build GRCh38
rs773677227 Chr.10: 132225304-132225304 on Build GRCh38
rs372579511 Chr.10: 132225380-132225380 on Build GRCh38
rs554190665 Chr.10: 132225424-132225424 on Build GRCh38
rs761415323 Chr.10: 132225463-132225463 on Build GRCh38
rs758349598 Chr.10: 132225410-132225410 on Build GRCh38
rs747087406 Chr.10: 132225483-132225483 on Build GRCh38
rs758093168 Chr.10: 132225322-132225322 on Build GRCh38
rs763813981 Chr.10: 132225387-132225387 on Build GRCh38
rs777029140 Chr.10: 132225419-132225419 on Build GRCh38
rs200904038 Chr.10: 132225333-132225333 on Build GRCh38
rs777240398 Chr.10: 132225328-132225328 on Build GRCh38
rs761112149 Chr.10: 132225374-132225374 on Build GRCh38
rs200405355 Chr.10: 132225378-132225378 on Build GRCh38

View Full Product Details