Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 17322406-17322665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHR
Entrez Gene ID
196
Gene Name
aryl hydrocarbon receptor
Gene Aliases
bHLHe76
UniGene
Hs.171189
Gene Chromosome Location
Chr.7: 17298652-17346151 on Build GRCh38
Transcripts
NM_001621.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4483643 c.271C>T p.P91S Chr.7: 17322518-17322518 on Build GRCh38
5141847 c.286G>A p.G96R Chr.7: 17322533-17322533 on Build GRCh38
1088081 c.314A>C p.N105T Chr.7: 17322561-17322561 on Build GRCh38
6020470 c.352T>A p.L118I Chr.7: 17322599-17322599 on Build GRCh38
4660607 c.360G>T p.Q120H Chr.7: 17322607-17322607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778158856 Chr.7: 17322468-17322468 on Build GRCh38
rs761566317 Chr.7: 17322532-17322532 on Build GRCh38
rs769439092 Chr.7: 17322577-17322577 on Build GRCh38
rs199498192 Chr.7: 17322573-17322573 on Build GRCh38
rs774150309 Chr.7: 17322522-17322522 on Build GRCh38
rs759677052 Chr.7: 17322542-17322542 on Build GRCh38
rs779560588 Chr.7: 17322458-17322458 on Build GRCh38
rs780162475 Chr.7: 17322600-17322601 on Build GRCh38
rs1050798246 Chr.7: 17322593-17322593 on Build GRCh38
rs771821651 Chr.7: 17322609-17322609 on Build GRCh38
rs752852966 Chr.7: 17322548-17322548 on Build GRCh38
rs1033678707 Chr.7: 17322444-17322444 on Build GRCh38
rs774406536 Chr.7: 17322543-17322543 on Build GRCh38
rs780743731 Chr.7: 17322503-17322503 on Build GRCh38
rs190138857 Chr.7: 17322518-17322518 on Build GRCh38
rs754076368 Chr.7: 17322535-17322535 on Build GRCh38
rs537938613 Chr.7: 17322582-17322582 on Build GRCh38
rs750248203 Chr.7: 17322487-17322488 on Build GRCh38
rs538650296 Chr.7: 17322440-17322440 on Build GRCh38
rs770595590 Chr.7: 17322477-17322477 on Build GRCh38
rs768620242 Chr.7: 17322597-17322597 on Build GRCh38
rs766592204 Chr.7: 17322533-17322533 on Build GRCh38
rs756059157 Chr.7: 17322597-17322598 on Build GRCh38
rs200324493 Chr.7: 17322551-17322551 on Build GRCh38
rs377724396 Chr.7: 17322559-17322559 on Build GRCh38
rs747355456 Chr.7: 17322476-17322476 on Build GRCh38
rs774992729 Chr.7: 17322516-17322516 on Build GRCh38
rs758977052 Chr.7: 17322463-17322463 on Build GRCh38
rs570282832 Chr.7: 17322554-17322554 on Build GRCh38
rs374791470 Chr.7: 17322586-17322586 on Build GRCh38
rs747971675 Chr.7: 17322606-17322606 on Build GRCh38
rs1002107456 Chr.7: 17322443-17322443 on Build GRCh38
rs375683016 Chr.7: 17322488-17322488 on Build GRCh38
rs150911949 Chr.7: 17322567-17322567 on Build GRCh38
rs759790577 Chr.7: 17322623-17322623 on Build GRCh38
rs1009297239 Chr.7: 17322625-17322625 on Build GRCh38
rs780881089 Chr.7: 17322570-17322570 on Build GRCh38
rs762813561 Chr.7: 17322517-17322517 on Build GRCh38
rs773075486 Chr.7: 17322611-17322611 on Build GRCh38
rs141518984 Chr.7: 17322505-17322505 on Build GRCh38
rs753316268 Chr.7: 17322462-17322462 on Build GRCh38
rs903910826 Chr.7: 17322479-17322479 on Build GRCh38
rs774195738 Chr.7: 17322602-17322602 on Build GRCh38
rs748786808 Chr.7: 17322498-17322498 on Build GRCh38
rs779501119 Chr.7: 17322583-17322583 on Build GRCh38

View Full Product Details