Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18618080-18618329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM59L
Entrez Gene ID
25789
Gene Name
transmembrane protein 59 like
Gene Aliases
BSMAP, C19orf4
UniGene
Hs.329850
Gene Chromosome Location
Chr.19: 18612872-18621039 on Build GRCh38
Transcripts
NM_012109.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7129931 c.768C>T p.L256L Chr.19: 18618258-18618258 on Build GRCh38
438978 c.740C>T p.S247F Chr.19: 18618230-18618230 on Build GRCh38
3100253 c.673G>T p.G225C Chr.19: 18618163-18618163 on Build GRCh38
6003550 c.717C>A p.T239T Chr.19: 18618207-18618207 on Build GRCh38
3783206 c.779C>T p.S260F Chr.19: 18618269-18618269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775192187 Chr.19: 18618228-18618228 on Build GRCh38
rs200853614 Chr.19: 18618225-18618225 on Build GRCh38
rs750535842 Chr.19: 18618178-18618178 on Build GRCh38
rs368780170 Chr.19: 18618130-18618130 on Build GRCh38
rs141763263 Chr.19: 18618197-18618197 on Build GRCh38
rs751114793 Chr.19: 18618147-18618148 on Build GRCh38
rs553416240 Chr.19: 18618272-18618272 on Build GRCh38
rs747912338 Chr.19: 18618108-18618108 on Build GRCh38
rs758134189 Chr.19: 18618206-18618206 on Build GRCh38
rs112435133 Chr.19: 18618299-18618300 on Build GRCh38
rs557065231 Chr.19: 18618203-18618203 on Build GRCh38
rs879026571 Chr.19: 18618299-18618299 on Build GRCh38
rs111685752 Chr.19: 18618145-18618145 on Build GRCh38
rs749901576 Chr.19: 18618295-18618296 on Build GRCh38
rs201231250 Chr.19: 18618218-18618218 on Build GRCh38
rs767222583 Chr.19: 18618289-18618290 on Build GRCh38
rs147406958 Chr.19: 18618158-18618158 on Build GRCh38
rs369305644 Chr.19: 18618121-18618121 on Build GRCh38
rs982613965 Chr.19: 18618250-18618250 on Build GRCh38
rs369691709 Chr.19: 18618304-18618304 on Build GRCh38
rs761889402 Chr.19: 18618290-18618290 on Build GRCh38
rs970717176 Chr.19: 18618245-18618245 on Build GRCh38
rs776180657 Chr.19: 18618144-18618144 on Build GRCh38
rs746680580 Chr.19: 18618127-18618127 on Build GRCh38
rs769254521 Chr.19: 18618266-18618266 on Build GRCh38
rs766463877 Chr.19: 18618302-18618302 on Build GRCh38
rs776196824 Chr.19: 18618238-18618238 on Build GRCh38
rs756194015 Chr.19: 18618188-18618188 on Build GRCh38
rs112872200 Chr.19: 18618300-18618300 on Build GRCh38
rs757503566 Chr.19: 18618157-18618157 on Build GRCh38
rs886953812 Chr.19: 18618196-18618196 on Build GRCh38
rs767754864 Chr.19: 18618295-18618295 on Build GRCh38
rs772842044 Chr.19: 18618125-18618125 on Build GRCh38
rs767453157 Chr.19: 18618172-18618172 on Build GRCh38
rs200281945 Chr.19: 18618271-18618271 on Build GRCh38
rs1024097296 Chr.19: 18618236-18618236 on Build GRCh38
rs199879621 Chr.19: 18618148-18618148 on Build GRCh38
rs199820188 Chr.19: 18618276-18618276 on Build GRCh38
rs201427140 Chr.19: 18618301-18618301 on Build GRCh38
rs1025357287 Chr.19: 18618164-18618164 on Build GRCh38
rs774885941 Chr.19: 18618289-18618289 on Build GRCh38
rs776238637 Chr.19: 18618131-18618131 on Build GRCh38
rs759913202 Chr.19: 18618134-18618134 on Build GRCh38
rs1039808881 Chr.19: 18618101-18618101 on Build GRCh38
rs745353654 Chr.19: 18618247-18618247 on Build GRCh38
rs765754270 Chr.19: 18618143-18618143 on Build GRCh38
rs779803230 Chr.19: 18618300-18618300 on Build GRCh38
rs746627149 Chr.19: 18618217-18618217 on Build GRCh38
rs777551956 Chr.19: 18618209-18618209 on Build GRCh38
rs761440020 Chr.19: 18618160-18618161 on Build GRCh38
rs1017499380 Chr.19: 18618305-18618305 on Build GRCh38

View Full Product Details