Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44776578-44776839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS8
Entrez Gene ID
6202
Gene Name
ribosomal protein S8
Gene Aliases
S8
UniGene
Hs.512675
Gene Chromosome Location
Chr.1: 44775574-44778740 on Build GRCh38
Transcripts
NM_001012.1 NR_000024.2

Gene Symbol
SNORD46
Entrez Gene ID
94161
Gene Name
small nucleolar RNA, C/D box 46
Gene Aliases
RNU40, RNU46, U40, U46
UniGene
Gene Chromosome Location
Chr.1: 44776492-44776589 on Build GRCh38
Transcripts
NM_001012.1 NR_000024.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195172 c.136G>A p.V46I Chr.1: 44776699-44776699 on Build GRCh38
6294044 c.145C>T p.R49W Chr.1: 44776708-44776708 on Build GRCh38
681210 c.146G>T p.R49L Chr.1: 44776709-44776709 on Build GRCh38
4805271 c.146G>C p.R49P Chr.1: 44776709-44776709 on Build GRCh38
910059 c.132C>T p.H44H Chr.1: 44776695-44776695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112943412 Chr.1: 44776679-44776679 on Build GRCh38
rs775697921 Chr.1: 44776795-44776795 on Build GRCh38
rs761409147 Chr.1: 44776741-44776741 on Build GRCh38
rs756916488 Chr.1: 44776637-44776637 on Build GRCh38
rs753641869 Chr.1: 44776729-44776729 on Build GRCh38
rs749918432 Chr.1: 44776747-44776747 on Build GRCh38
rs368998223 Chr.1: 44776770-44776770 on Build GRCh38
rs756591403 Chr.1: 44776774-44776774 on Build GRCh38
rs767523711 Chr.1: 44776618-44776618 on Build GRCh38
rs757839585 Chr.1: 44776749-44776749 on Build GRCh38
rs755507477 Chr.1: 44776653-44776653 on Build GRCh38
rs748715739 Chr.1: 44776653-44776653 on Build GRCh38
rs755914026 Chr.1: 44776632-44776632 on Build GRCh38
rs189273957 Chr.1: 44776664-44776664 on Build GRCh38
rs563761308 Chr.1: 44776661-44776661 on Build GRCh38
rs771153794 Chr.1: 44776688-44776688 on Build GRCh38
rs373290917 Chr.1: 44776633-44776633 on Build GRCh38
rs903464767 Chr.1: 44776663-44776663 on Build GRCh38
rs745944753 Chr.1: 44776792-44776792 on Build GRCh38
rs773617525 Chr.1: 44776672-44776672 on Build GRCh38
rs753499357 Chr.1: 44776636-44776636 on Build GRCh38
rs779137077 Chr.1: 44776791-44776791 on Build GRCh38
rs759612522 Chr.1: 44776699-44776699 on Build GRCh38
rs184816863 Chr.1: 44776628-44776628 on Build GRCh38
rs201440654 Chr.1: 44776651-44776651 on Build GRCh38
rs779539651 Chr.1: 44776761-44776761 on Build GRCh38
rs778332041 Chr.1: 44776639-44776639 on Build GRCh38
rs112706003 Chr.1: 44776777-44776777 on Build GRCh38
rs774468443 Chr.1: 44776610-44776610 on Build GRCh38
rs147164346 Chr.1: 44776708-44776708 on Build GRCh38
rs115967171 Chr.1: 44776794-44776794 on Build GRCh38
rs760685360 Chr.1: 44776717-44776717 on Build GRCh38
rs1021784685 Chr.1: 44776638-44776638 on Build GRCh38
rs772147655 Chr.1: 44776701-44776701 on Build GRCh38
rs370695473 Chr.1: 44776616-44776616 on Build GRCh38
rs759433169 Chr.1: 44776614-44776614 on Build GRCh38
rs760740489 Chr.1: 44776806-44776806 on Build GRCh38
rs999504137 Chr.1: 44776787-44776787 on Build GRCh38
rs373420727 Chr.1: 44776781-44776781 on Build GRCh38
rs749523000 Chr.1: 44776675-44776675 on Build GRCh38
rs771081823 Chr.1: 44776785-44776785 on Build GRCh38
rs745349849 Chr.1: 44776647-44776647 on Build GRCh38
rs766427693 Chr.1: 44776609-44776609 on Build GRCh38
rs377275681 Chr.1: 44776624-44776624 on Build GRCh38
rs774328393 Chr.1: 44776698-44776698 on Build GRCh38
rs776310845 Chr.1: 44776648-44776648 on Build GRCh38
rs552685243 Chr.1: 44776746-44776746 on Build GRCh38
rs763235891 Chr.1: 44776601-44776601 on Build GRCh38
rs376448784 Chr.1: 44776793-44776793 on Build GRCh38
rs764035373 Chr.1: 44776725-44776725 on Build GRCh38

View Full Product Details