Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136875183-136875426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBMX
Entrez Gene ID
27316
Gene Name
RNA binding motif protein, X-linked
Gene Aliases
HNRNPG, HNRPG, MRXS11, RBMXP1, RBMXRT, RNMX, hnRNP-G
UniGene
Hs.380118
Gene Chromosome Location
Chr.X: 136869194-136880780 on Build GRCh38
Transcripts
NM_001164803.1 NM_002139.3 NR_028476.1 NR_028477.1

SNP Detail

SNP ID Chromosome Location
rs750130071 Chr.X: 136875293-136875293 on Build GRCh38
rs375183129 Chr.X: 136875222-136875222 on Build GRCh38
rs368183974 Chr.X: 136875272-136875272 on Build GRCh38
rs748551065 Chr.X: 136875348-136875348 on Build GRCh38
rs770604892 Chr.X: 136875394-136875395 on Build GRCh38
rs779596723 Chr.X: 136875318-136875318 on Build GRCh38
rs747307792 Chr.X: 136875375-136875375 on Build GRCh38
rs28595841 Chr.X: 136875326-136875326 on Build GRCh38
rs760772574 Chr.X: 136875215-136875215 on Build GRCh38
rs772520857 Chr.X: 136875355-136875355 on Build GRCh38
rs767168915 Chr.X: 136875276-136875276 on Build GRCh38
rs769109014 Chr.X: 136875317-136875317 on Build GRCh38
rs771048795 Chr.X: 136875394-136875394 on Build GRCh38
rs548166441 Chr.X: 136875252-136875252 on Build GRCh38
rs776833261 Chr.X: 136875230-136875230 on Build GRCh38
rs773501418 Chr.X: 136875213-136875213 on Build GRCh38
rs777163606 Chr.X: 136875327-136875327 on Build GRCh38
rs755698474 Chr.X: 136875294-136875294 on Build GRCh38
rs781262836 Chr.X: 136875287-136875287 on Build GRCh38
rs891396849 Chr.X: 136875290-136875290 on Build GRCh38
rs765121298 Chr.X: 136875393-136875394 on Build GRCh38
rs1004843607 Chr.X: 136875291-136875291 on Build GRCh38
rs3814292 Chr.X: 136875371-136875371 on Build GRCh38
rs2306221 Chr.X: 136875219-136875219 on Build GRCh38
rs201924324 Chr.X: 136875334-136875334 on Build GRCh38
rs763044365 Chr.X: 136875241-136875241 on Build GRCh38
rs771667003 Chr.X: 136875246-136875247 on Build GRCh38
rs375856058 Chr.X: 136875379-136875379 on Build GRCh38
rs377120509 Chr.X: 136875342-136875342 on Build GRCh38
rs11556431 Chr.X: 136875288-136875288 on Build GRCh38
rs978869966 Chr.X: 136875231-136875231 on Build GRCh38
rs922110575 Chr.X: 136875278-136875278 on Build GRCh38
rs184511527 Chr.X: 136875260-136875260 on Build GRCh38
rs772609375 Chr.X: 136875393-136875393 on Build GRCh38
rs764123593 Chr.X: 136875244-136875244 on Build GRCh38

View Full Product Details