Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 161420093-161420364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBR1
Entrez Gene ID
10716
Gene Name
T-box, brain 1
Gene Aliases
TBR-1, TES-56
UniGene
Hs.210862
Gene Chromosome Location
Chr.2: 161416094-161425867 on Build GRCh38
Transcripts
NM_006593.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4732496 c.1172T>G p.F391C Chr.2: 161420239-161420239 on Build GRCh38
4933278 c.1166A>T p.K389I Chr.2: 161420233-161420233 on Build GRCh38
6797828 c.1190+1G>A p.? Chr.2: 161420258-161420258 on Build GRCh38
3708982 c.1140G>T p.L380L Chr.2: 161420207-161420207 on Build GRCh38
1400409 c.1174C>T p.R392W Chr.2: 161420241-161420241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181943543 Chr.2: 161420228-161420228 on Build GRCh38
rs549284676 Chr.2: 161420162-161420162 on Build GRCh38
rs997973150 Chr.2: 161420335-161420335 on Build GRCh38
rs200776596 Chr.2: 161420192-161420192 on Build GRCh38
rs1036197838 Chr.2: 161420336-161420337 on Build GRCh38
rs762713626 Chr.2: 161420222-161420222 on Build GRCh38
rs79294493 Chr.2: 161420201-161420201 on Build GRCh38
rs746242590 Chr.2: 161420155-161420155 on Build GRCh38
rs189181205 Chr.2: 161420213-161420213 on Build GRCh38
rs756985721 Chr.2: 161420179-161420179 on Build GRCh38
rs778781183 Chr.2: 161420154-161420154 on Build GRCh38
rs1056495412 Chr.2: 161420315-161420315 on Build GRCh38
rs753542347 Chr.2: 161420178-161420179 on Build GRCh38
rs768211070 Chr.2: 161420165-161420165 on Build GRCh38
rs772779182 Chr.2: 161420217-161420217 on Build GRCh38
rs375081745 Chr.2: 161420300-161420300 on Build GRCh38
rs1036311270 Chr.2: 161420130-161420130 on Build GRCh38
rs879854659 Chr.2: 161420146-161420146 on Build GRCh38
rs764023842 Chr.2: 161420297-161420297 on Build GRCh38
rs766973299 Chr.2: 161420324-161420324 on Build GRCh38
rs759476148 Chr.2: 161420271-161420271 on Build GRCh38
rs780655123 Chr.2: 161420159-161420159 on Build GRCh38
rs774232975 Chr.2: 161420270-161420270 on Build GRCh38
rs369968271 Chr.2: 161420309-161420309 on Build GRCh38
rs911535184 Chr.2: 161420121-161420121 on Build GRCh38
rs371795758 Chr.2: 161420149-161420149 on Build GRCh38
rs200973809 Chr.2: 161420173-161420173 on Build GRCh38
rs758878063 Chr.2: 161420156-161420156 on Build GRCh38
rs753942414 Chr.2: 161420145-161420145 on Build GRCh38

View Full Product Details