Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1535494-1535745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM240
Entrez Gene ID
339453
Gene Name
transmembrane protein 240
Gene Aliases
C1orf70, SCA21
UniGene
Hs.668654
Gene Chromosome Location
Chr.1: 1534778-1540360 on Build GRCh38
Transcripts
NM_001114748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2121781 c.343G>A p.V115M Chr.1: 1535619-1535619 on Build GRCh38
7214547 c.374-5C>T p.? Chr.1: 1535512-1535512 on Build GRCh38
1333701 c.352T>A p.W118R Chr.1: 1535610-1535610 on Build GRCh38
6335288 c.282_286delGCTGC p.L95fs*>78 Chr.1: 1535676-1535680 on Build GRCh38
1333703 c.286C>T p.L96L Chr.1: 1535676-1535676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568810688 Chr.1: 1535532-1535532 on Build GRCh38
rs762787977 Chr.1: 1535697-1535697 on Build GRCh38
rs183867156 Chr.1: 1535602-1535602 on Build GRCh38
rs758010368 Chr.1: 1535515-1535515 on Build GRCh38
rs964847659 Chr.1: 1535671-1535671 on Build GRCh38
rs778858593 Chr.1: 1535511-1535511 on Build GRCh38
rs994275115 Chr.1: 1535549-1535549 on Build GRCh38
rs376390462 Chr.1: 1535666-1535666 on Build GRCh38
rs752556739 Chr.1: 1535572-1535572 on Build GRCh38
rs746611425 Chr.1: 1535517-1535517 on Build GRCh38
rs763953379 Chr.1: 1535698-1535698 on Build GRCh38
rs554736432 Chr.1: 1535563-1535563 on Build GRCh38
rs765411959 Chr.1: 1535578-1535580 on Build GRCh38
rs867534273 Chr.1: 1535634-1535634 on Build GRCh38
rs606231453 Chr.1: 1535616-1535616 on Build GRCh38
rs769269506 Chr.1: 1535613-1535613 on Build GRCh38
rs376315923 Chr.1: 1535635-1535635 on Build GRCh38
rs749750055 Chr.1: 1535615-1535615 on Build GRCh38
rs936719939 Chr.1: 1535593-1535593 on Build GRCh38
rs542091821 Chr.1: 1535623-1535623 on Build GRCh38
rs977231573 Chr.1: 1535520-1535520 on Build GRCh38
rs540694977 Chr.1: 1535577-1535577 on Build GRCh38
rs1055219640 Chr.1: 1535595-1535595 on Build GRCh38
rs770244919 Chr.1: 1535543-1535543 on Build GRCh38
rs752708373 Chr.1: 1535701-1535703 on Build GRCh38
rs546208867 Chr.1: 1535601-1535601 on Build GRCh38
rs758255905 Chr.1: 1535574-1535574 on Build GRCh38
rs774697071 Chr.1: 1535550-1535550 on Build GRCh38
rs1106226 Chr.1: 1535512-1535512 on Build GRCh38
rs187039783 Chr.1: 1535637-1535637 on Build GRCh38
rs948435492 Chr.1: 1535618-1535618 on Build GRCh38
rs765175519 Chr.1: 1535571-1535571 on Build GRCh38
rs373327807 Chr.1: 1535649-1535649 on Build GRCh38
rs780648238 Chr.1: 1535614-1535614 on Build GRCh38
rs975766887 Chr.1: 1535517-1535518 on Build GRCh38
rs577269741 Chr.1: 1535598-1535598 on Build GRCh38
rs967093969 Chr.1: 1535714-1535714 on Build GRCh38
rs606231454 Chr.1: 1535723-1535723 on Build GRCh38
rs964726593 Chr.1: 1535516-1535516 on Build GRCh38
rs775738452 Chr.1: 1535544-1535544 on Build GRCh38
rs762110410 Chr.1: 1535552-1535552 on Build GRCh38
rs537680601 Chr.1: 1535537-1535537 on Build GRCh38
rs553883626 Chr.1: 1535589-1535589 on Build GRCh38
rs966591411 Chr.1: 1535718-1535718 on Build GRCh38
rs532021282 Chr.1: 1535620-1535620 on Build GRCh38
rs750465095 Chr.1: 1535565-1535565 on Build GRCh38
rs773451198 Chr.1: 1535638-1535638 on Build GRCh38
rs769114802 Chr.1: 1535674-1535674 on Build GRCh38
rs763420851 Chr.1: 1535545-1535545 on Build GRCh38
rs777426659 Chr.1: 1535578-1535578 on Build GRCh38
rs755164772 Chr.1: 1535520-1535521 on Build GRCh38
rs371009717 Chr.1: 1535722-1535722 on Build GRCh38
rs994122912 Chr.1: 1535564-1535564 on Build GRCh38
rs747363918 Chr.1: 1535546-1535546 on Build GRCh38
rs770496430 Chr.1: 1535525-1535525 on Build GRCh38
rs1045410944 Chr.1: 1535619-1535619 on Build GRCh38
rs997999010 Chr.1: 1535673-1535673 on Build GRCh38
rs924437505 Chr.1: 1535531-1535531 on Build GRCh38
rs951857470 Chr.1: 1535558-1535558 on Build GRCh38
rs574855348 Chr.1: 1535569-1535569 on Build GRCh38
rs760887921 Chr.1: 1535653-1535653 on Build GRCh38
rs555582754 Chr.1: 1535590-1535590 on Build GRCh38
rs780622089 Chr.1: 1535533-1535533 on Build GRCh38
rs977896123 Chr.1: 1535586-1535586 on Build GRCh38

View Full Product Details