Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37637427-37637680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC4
Entrez Gene ID
7223
Gene Name
transient receptor potential cation channel subfamily C member 4
Gene Aliases
HTRP-4, HTRP4, TRP4
UniGene
Hs.262960
Gene Chromosome Location
Chr.13: 37633872-37870374 on Build GRCh38
Transcripts
NM_001135955.1 NM_001135956.1 NM_001135957.1 NM_001135958.1 NM_003306.1 NM_016179.2 XM_011535206.1 XM_017020723.1 XM_017020724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947222 c.2304G>A p.S768S Chr.13: 37637533-37637533 on Build GRCh38
219476 c.2249T>A p.V750D Chr.13: 37637588-37637588 on Build GRCh38
947218 c.2376G>T p.K792N Chr.13: 37637461-37637461 on Build GRCh38
6809242 c.2339_2340insA p.S781fs*2 Chr.13: 37637497-37637498 on Build GRCh38
4047477 c.2247A>C p.E749D Chr.13: 37637590-37637590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112903780 Chr.13: 37637630-37637630 on Build GRCh38
rs773477061 Chr.13: 37637563-37637563 on Build GRCh38
rs754344232 Chr.13: 37637509-37637509 on Build GRCh38
rs146807006 Chr.13: 37637565-37637565 on Build GRCh38
rs754182455 Chr.13: 37637581-37637581 on Build GRCh38
rs751284923 Chr.13: 37637638-37637638 on Build GRCh38
rs758310603 Chr.13: 37637557-37637557 on Build GRCh38
rs377554360 Chr.13: 37637579-37637579 on Build GRCh38
rs762033326 Chr.13: 37637471-37637471 on Build GRCh38
rs1055947215 Chr.13: 37637543-37637543 on Build GRCh38
rs776480841 Chr.13: 37637461-37637461 on Build GRCh38
rs758030354 Chr.13: 37637631-37637631 on Build GRCh38
rs761951483 Chr.13: 37637559-37637559 on Build GRCh38
rs759483362 Chr.13: 37637462-37637462 on Build GRCh38
rs772127889 Chr.13: 37637562-37637562 on Build GRCh38
rs561992636 Chr.13: 37637512-37637512 on Build GRCh38
rs774437900 Chr.13: 37637469-37637469 on Build GRCh38
rs767663885 Chr.13: 37637477-37637477 on Build GRCh38
rs947179279 Chr.13: 37637614-37637614 on Build GRCh38
rs759976587 Chr.13: 37637597-37637597 on Build GRCh38
rs746638345 Chr.13: 37637528-37637528 on Build GRCh38
rs774295967 Chr.13: 37637558-37637558 on Build GRCh38
rs766747428 Chr.13: 37637495-37637495 on Build GRCh38
rs186177283 Chr.13: 37637516-37637516 on Build GRCh38
rs750641378 Chr.13: 37637484-37637484 on Build GRCh38
rs140911831 Chr.13: 37637534-37637534 on Build GRCh38
rs765838206 Chr.13: 37637598-37637598 on Build GRCh38
rs776443992 Chr.13: 37637542-37637542 on Build GRCh38
rs762649694 Chr.13: 37637476-37637476 on Build GRCh38
rs1046643529 Chr.13: 37637503-37637503 on Build GRCh38
rs745810881 Chr.13: 37637552-37637552 on Build GRCh38
rs756533828 Chr.13: 37637487-37637487 on Build GRCh38
rs756141698 Chr.13: 37637629-37637629 on Build GRCh38
rs143708525 Chr.13: 37637553-37637553 on Build GRCh38
rs767662289 Chr.13: 37637519-37637519 on Build GRCh38
rs755471814 Chr.13: 37637510-37637510 on Build GRCh38
rs764107733 Chr.13: 37637468-37637468 on Build GRCh38

View Full Product Details