Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52598557-52598798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP46
Entrez Gene ID
64854
Gene Name
ubiquitin specific peptidase 46
Gene Aliases
-
UniGene
Hs.7966
Gene Chromosome Location
Chr.4: 52590960-52659335 on Build GRCh38
Transcripts
NM_001134223.1 NM_001286767.1 NM_001286768.1 NM_022832.3 XM_017008557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1633712 c.949A>G p.I317V Chr.4: 52598678-52598678 on Build GRCh38
5866506 c.964G>A p.G322S Chr.4: 52598663-52598663 on Build GRCh38
6327108 c.943G>T p.G315C Chr.4: 52598663-52598663 on Build GRCh38
1430015 c.957A>C p.K319N Chr.4: 52598670-52598670 on Build GRCh38
5829931 c.936A>C p.K312N Chr.4: 52598670-52598670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771644189 Chr.4: 52598658-52598658 on Build GRCh38
rs756630799 Chr.4: 52598754-52598755 on Build GRCh38
rs868571577 Chr.4: 52598710-52598710 on Build GRCh38
rs578220961 Chr.4: 52598706-52598706 on Build GRCh38
rs749150595 Chr.4: 52598597-52598597 on Build GRCh38
rs759992397 Chr.4: 52598691-52598691 on Build GRCh38
rs778645321 Chr.4: 52598628-52598628 on Build GRCh38
rs553899320 Chr.4: 52598605-52598605 on Build GRCh38
rs927629569 Chr.4: 52598742-52598742 on Build GRCh38
rs757326314 Chr.4: 52598749-52598749 on Build GRCh38
rs374587502 Chr.4: 52598747-52598747 on Build GRCh38
rs772802080 Chr.4: 52598663-52598663 on Build GRCh38
rs377620169 Chr.4: 52598619-52598619 on Build GRCh38
rs184517651 Chr.4: 52598724-52598724 on Build GRCh38
rs779938763 Chr.4: 52598595-52598595 on Build GRCh38
rs17051590 Chr.4: 52598664-52598664 on Build GRCh38
rs766363845 Chr.4: 52598751-52598751 on Build GRCh38
rs775809348 Chr.4: 52598718-52598718 on Build GRCh38
rs558384288 Chr.4: 52598680-52598680 on Build GRCh38
rs747690016 Chr.4: 52598634-52598634 on Build GRCh38

View Full Product Details