Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 158879631-158879868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR60
Entrez Gene ID
55112
Gene Name
WD repeat domain 60
Gene Aliases
FAP163, SRPS6, SRTD8
UniGene
Hs.389945
Gene Chromosome Location
Chr.7: 158839250-158958695 on Build GRCh38
Transcripts
NM_018051.4 XM_005249549.1 XM_006716041.2 XM_011516367.1 XM_011516368.2 XM_011516369.2 XM_017012381.1 XM_017012382.1 XM_017012383.1 XM_017012384.1 XM_017012385.1 XM_017012386.1 XM_017012387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014779 c.721T>G p.S241A Chr.7: 158879831-158879831 on Build GRCh38
5780259 c.705A>T p.E235D Chr.7: 158879815-158879815 on Build GRCh38
3636874 c.584G>A p.G195E Chr.7: 158879694-158879694 on Build GRCh38
6515847 c.652C>A p.P218T Chr.7: 158879762-158879762 on Build GRCh38
6247235 c.576G>T p.L192L Chr.7: 158879686-158879686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746656659 Chr.7: 158879664-158879664 on Build GRCh38
rs760704544 Chr.7: 158879777-158879777 on Build GRCh38
rs201379330 Chr.7: 158879688-158879688 on Build GRCh38
rs773243945 Chr.7: 158879773-158879773 on Build GRCh38
rs769162738 Chr.7: 158879675-158879675 on Build GRCh38
rs199667516 Chr.7: 158879782-158879782 on Build GRCh38
rs754088031 Chr.7: 158879697-158879697 on Build GRCh38
rs754720217 Chr.7: 158879700-158879700 on Build GRCh38
rs778784309 Chr.7: 158879705-158879705 on Build GRCh38
rs200520038 Chr.7: 158879722-158879722 on Build GRCh38
rs760059618 Chr.7: 158879719-158879719 on Build GRCh38
rs770678221 Chr.7: 158879728-158879728 on Build GRCh38
rs375240436 Chr.7: 158879743-158879743 on Build GRCh38
rs774841970 Chr.7: 158879740-158879740 on Build GRCh38
rs766627549 Chr.7: 158879696-158879696 on Build GRCh38
rs764480798 Chr.7: 158879685-158879686 on Build GRCh38
rs768833360 Chr.7: 158879739-158879739 on Build GRCh38
rs577876445 Chr.7: 158879690-158879690 on Build GRCh38
rs562885560 Chr.7: 158879790-158879790 on Build GRCh38
rs1055788086 Chr.7: 158879717-158879717 on Build GRCh38
rs759760480 Chr.7: 158879788-158879788 on Build GRCh38
rs368847195 Chr.7: 158879692-158879692 on Build GRCh38
rs760864089 Chr.7: 158879693-158879693 on Build GRCh38
rs762148102 Chr.7: 158879801-158879802 on Build GRCh38
rs775978095 Chr.7: 158879673-158879673 on Build GRCh38
rs780955608 Chr.7: 158879731-158879731 on Build GRCh38
rs188181596 Chr.7: 158879674-158879674 on Build GRCh38
rs150548113 Chr.7: 158879824-158879824 on Build GRCh38
rs765728280 Chr.7: 158879735-158879735 on Build GRCh38
rs752510071 Chr.7: 158879709-158879709 on Build GRCh38
rs752596216 Chr.7: 158879792-158879792 on Build GRCh38
rs751913477 Chr.7: 158879781-158879781 on Build GRCh38
rs2709859 Chr.7: 158879785-158879785 on Build GRCh38
rs750519654 Chr.7: 158879830-158879830 on Build GRCh38
rs758249537 Chr.7: 158879793-158879793 on Build GRCh38
rs745507098 Chr.7: 158879833-158879833 on Build GRCh38
rs767577574 Chr.7: 158879819-158879820 on Build GRCh38
rs751581916 Chr.7: 158879805-158879805 on Build GRCh38
rs774660510 Chr.7: 158879682-158879682 on Build GRCh38
rs780851536 Chr.7: 158879825-158879825 on Build GRCh38
rs73527702 Chr.7: 158879714-158879714 on Build GRCh38
rs777808616 Chr.7: 158879803-158879803 on Build GRCh38
rs770768082 Chr.7: 158879667-158879667 on Build GRCh38
rs1035721899 Chr.7: 158879772-158879772 on Build GRCh38
rs1023573871 Chr.7: 158879726-158879726 on Build GRCh38
rs772683938 Chr.7: 158879744-158879744 on Build GRCh38

View Full Product Details