Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155842327-155842584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC33A1
Entrez Gene ID
9197
Gene Name
solute carrier family 33 member 1
Gene Aliases
ACATN, AT-1, AT1, CCHLND, SPG42
UniGene
Hs.478031
Gene Chromosome Location
Chr.3: 155826511-155854459 on Build GRCh38
Transcripts
NM_001190992.1 NM_004733.3 XM_011513311.2 XM_017007462.1 XM_017007463.1 XM_017007464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5647897 c.814A>G p.T272A Chr.3: 155842581-155842581 on Build GRCh38
80531 c.846C>T p.N282N Chr.3: 155842549-155842549 on Build GRCh38
3781103 c.819A>G p.T273T Chr.3: 155842576-155842576 on Build GRCh38
7319855 c.902T>C p.L301P Chr.3: 155842493-155842493 on Build GRCh38
7390443 c.868G>A p.E290K Chr.3: 155842527-155842527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377484298 Chr.3: 155842545-155842545 on Build GRCh38
rs577049995 Chr.3: 155842495-155842495 on Build GRCh38
rs371208781 Chr.3: 155842439-155842439 on Build GRCh38
rs1042717684 Chr.3: 155842492-155842492 on Build GRCh38
rs778986157 Chr.3: 155842556-155842562 on Build GRCh38
rs757509671 Chr.3: 155842545-155842547 on Build GRCh38
rs947109619 Chr.3: 155842398-155842398 on Build GRCh38
rs985901251 Chr.3: 155842373-155842373 on Build GRCh38
rs755080609 Chr.3: 155842563-155842563 on Build GRCh38
rs886058109 Chr.3: 155842491-155842491 on Build GRCh38
rs201312848 Chr.3: 155842408-155842408 on Build GRCh38
rs765511075 Chr.3: 155842536-155842536 on Build GRCh38
rs143409270 Chr.3: 155842516-155842516 on Build GRCh38
rs1011163387 Chr.3: 155842491-155842491 on Build GRCh38
rs531122837 Chr.3: 155842470-155842470 on Build GRCh38
rs182334558 Chr.3: 155842540-155842540 on Build GRCh38
rs146695209 Chr.3: 155842521-155842521 on Build GRCh38
rs770599492 Chr.3: 155842395-155842395 on Build GRCh38
rs766887166 Chr.3: 155842546-155842546 on Build GRCh38
rs200789237 Chr.3: 155842548-155842548 on Build GRCh38
rs368778917 Chr.3: 155842396-155842396 on Build GRCh38
rs373761577 Chr.3: 155842388-155842388 on Build GRCh38
rs558773904 Chr.3: 155842447-155842447 on Build GRCh38
rs749069368 Chr.3: 155842392-155842392 on Build GRCh38

View Full Product Details