Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73903901-73904170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF410
Entrez Gene ID
57862
Gene Name
zinc finger protein 410
Gene Aliases
APA-1, APA1
UniGene
Hs.270869
Gene Chromosome Location
Chr.14: 73886615-73932288 on Build GRCh38
Transcripts
NM_001242924.1 NM_001242926.1 NM_001242927.1 NM_001242928.1 NM_021188.2 NR_040251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4623017 c.626C>A p.S209Y Chr.14: 73904005-73904005 on Build GRCh38
3724783 c.691G>A p.V231I Chr.14: 73904070-73904070 on Build GRCh38
698698 c.670G>C p.E224Q Chr.14: 73904049-73904049 on Build GRCh38
6015574 c.731+2T>A p.? Chr.14: 73904112-73904112 on Build GRCh38
1128202 c.583G>A p.E195K Chr.14: 73903962-73903962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754420071 Chr.14: 73904131-73904131 on Build GRCh38
rs747784196 Chr.14: 73904142-73904142 on Build GRCh38
rs778435064 Chr.14: 73904137-73904137 on Build GRCh38
rs866815280 Chr.14: 73904109-73904109 on Build GRCh38
rs759607124 Chr.14: 73904017-73904017 on Build GRCh38
rs1013828980 Chr.14: 73904059-73904059 on Build GRCh38
rs776748874 Chr.14: 73904011-73904011 on Build GRCh38
rs748753941 Chr.14: 73903969-73903969 on Build GRCh38
rs753435028 Chr.14: 73904123-73904123 on Build GRCh38
rs757856614 Chr.14: 73904110-73904110 on Build GRCh38
rs936126640 Chr.14: 73904003-73904003 on Build GRCh38
rs374718824 Chr.14: 73904047-73904047 on Build GRCh38
rs374333979 Chr.14: 73903934-73903934 on Build GRCh38
rs925936596 Chr.14: 73903990-73903990 on Build GRCh38
rs547911591 Chr.14: 73904144-73904144 on Build GRCh38
rs781762161 Chr.14: 73904122-73904122 on Build GRCh38
rs778395550 Chr.14: 73903978-73903978 on Build GRCh38
rs748020664 Chr.14: 73903997-73903997 on Build GRCh38
rs751908500 Chr.14: 73904084-73904084 on Build GRCh38
rs948964914 Chr.14: 73903964-73903964 on Build GRCh38
rs1044413821 Chr.14: 73903966-73903966 on Build GRCh38
rs368479059 Chr.14: 73903940-73903940 on Build GRCh38
rs372362861 Chr.14: 73904036-73904036 on Build GRCh38
rs772949144 Chr.14: 73904006-73904006 on Build GRCh38
rs554869233 Chr.14: 73904010-73904010 on Build GRCh38
rs891853400 Chr.14: 73904061-73904061 on Build GRCh38
rs748195114 Chr.14: 73904082-73904082 on Build GRCh38
rs768415927 Chr.14: 73903972-73903972 on Build GRCh38
rs779718414 Chr.14: 73903957-73903957 on Build GRCh38
rs771782919 Chr.14: 73903998-73903998 on Build GRCh38
rs764400438 Chr.14: 73904062-73904062 on Build GRCh38
rs112979968 Chr.14: 73904015-73904015 on Build GRCh38
rs760608853 Chr.14: 73904009-73904009 on Build GRCh38

View Full Product Details