Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 94653183-94653442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLF1
Entrez Gene ID
84250
Gene Name
SMC5-SMC6 complex localization factor 1
Gene Aliases
ANKRD32, BRCTD1, BRCTx
UniGene
Hs.657315
Gene Chromosome Location
Chr.5: 94613477-94697733 on Build GRCh38
Transcripts
NM_032290.3 XM_017009978.1 XM_017009979.1 XM_017009980.1 XM_017009981.1 XM_017009982.1 XM_017009984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6371656 c.985G>A p.E329K Chr.5: 94653374-94653374 on Build GRCh38
1439205 c.1011G>T p.R337S Chr.5: 94653400-94653400 on Build GRCh38
4160061 c.891T>A p.I297I Chr.5: 94653280-94653280 on Build GRCh38
4160059 c.886G>A p.E296K Chr.5: 94653275-94653275 on Build GRCh38
4160060 c.890T>G p.I297S Chr.5: 94653279-94653279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961075235 Chr.5: 94653225-94653225 on Build GRCh38
rs765926259 Chr.5: 94653364-94653364 on Build GRCh38
rs201586187 Chr.5: 94653344-94653344 on Build GRCh38
rs1042272284 Chr.5: 94653372-94653372 on Build GRCh38
rs561197322 Chr.5: 94653280-94653280 on Build GRCh38
rs769960353 Chr.5: 94653233-94653233 on Build GRCh38
rs906959873 Chr.5: 94653400-94653400 on Build GRCh38
rs749606875 Chr.5: 94653266-94653266 on Build GRCh38
rs142724600 Chr.5: 94653286-94653286 on Build GRCh38
rs372908443 Chr.5: 94653297-94653297 on Build GRCh38
rs772730506 Chr.5: 94653314-94653314 on Build GRCh38
rs573549884 Chr.5: 94653265-94653265 on Build GRCh38
rs745966719 Chr.5: 94653226-94653226 on Build GRCh38
rs774323346 Chr.5: 94653289-94653290 on Build GRCh38
rs778747993 Chr.5: 94653235-94653235 on Build GRCh38
rs896494573 Chr.5: 94653365-94653365 on Build GRCh38
rs371383467 Chr.5: 94653228-94653228 on Build GRCh38
rs938007512 Chr.5: 94653329-94653329 on Build GRCh38
rs540954203 Chr.5: 94653221-94653221 on Build GRCh38
rs760301345 Chr.5: 94653354-94653354 on Build GRCh38
rs1009573549 Chr.5: 94653402-94653402 on Build GRCh38

View Full Product Details