Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18865879-18866134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPSAP2
Entrez Gene ID
5636
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 2
Gene Aliases
PAP41
UniGene
Hs.632236
Gene Chromosome Location
Chr.17: 18856299-18931287 on Build GRCh38
Transcripts
NM_001243936.1 NM_001243940.1 NM_001243941.1 NM_001243942.1 NM_002767.3 XM_005256725.2 XM_005256726.1 XM_005256729.4 XM_017024864.1 XM_017024865.1 XM_017024866.1 XM_017024867.1 XM_017024868.1 XM_017024869.1 XM_017024870.1 XM_017024871.1 XM_017024872.1 XM_017024873.1 XM_017024874.1 XM_017024875.1 XM_017024876.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347879 c.106A>C p.K36Q Chr.17: 18865939-18865939 on Build GRCh38
1324120 c.88T>G p.S30A Chr.17: 18865921-18865921 on Build GRCh38
3514989 c.48C>T p.T16T Chr.17: 18865881-18865881 on Build GRCh38
3514990 c.95T>C p.M32T Chr.17: 18865928-18865928 on Build GRCh38
976359 c.108G>T p.K36N Chr.17: 18865941-18865941 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs2386404 Chr.17: 18866036-18866036 on Build GRCh38
rs764462595 Chr.17: 18865960-18865962 on Build GRCh38
rs751673894 Chr.17: 18865908-18865908 on Build GRCh38
rs777506898 Chr.17: 18865987-18865987 on Build GRCh38
rs760809897 Chr.17: 18865930-18865931 on Build GRCh38
rs545838819 Chr.17: 18866094-18866094 on Build GRCh38
rs756882819 Chr.17: 18865993-18865993 on Build GRCh38
rs533463778 Chr.17: 18866017-18866017 on Build GRCh38
rs199793178 Chr.17: 18865931-18865931 on Build GRCh38
rs559554657 Chr.17: 18866100-18866100 on Build GRCh38
rs531524791 Chr.17: 18866094-18866095 on Build GRCh38
rs925201158 Chr.17: 18866093-18866094 on Build GRCh38
rs866413174 Chr.17: 18865929-18865930 on Build GRCh38
rs35736659 Chr.17: 18866007-18866008 on Build GRCh38
rs868518153 Chr.17: 18865913-18865913 on Build GRCh38
rs543700218 Chr.17: 18865930-18865930 on Build GRCh38
rs563389852 Chr.17: 18866016-18866016 on Build GRCh38
rs755964061 Chr.17: 18865975-18865975 on Build GRCh38
rs977441254 Chr.17: 18866076-18866094 on Build GRCh38
rs754933880 Chr.17: 18865925-18865925 on Build GRCh38
rs374005937 Chr.17: 18865914-18865914 on Build GRCh38
rs111752362 Chr.17: 18866047-18866047 on Build GRCh38
rs112112885 Chr.17: 18866052-18866052 on Build GRCh38
rs568339974 Chr.17: 18866101-18866102 on Build GRCh38
rs368913854 Chr.17: 18865902-18865902 on Build GRCh38
rs992412062 Chr.17: 18866000-18866000 on Build GRCh38
rs1049759183 Chr.17: 18866009-18866009 on Build GRCh38

View Full Product Details