Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131478053-131478290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MZT2A
Entrez Gene ID
653784
Gene Name
mitotic spindle organizing protein 2A
Gene Aliases
FAM128A, MOZART2A
UniGene
Hs.655067
Gene Chromosome Location
Chr.2: 131469724-131492719 on Build GRCh38
Transcripts
XM_005263742.2 NM_080386.3

Gene Symbol
TUBA3D
Entrez Gene ID
113457
Gene Name
tubulin alpha 3d
Gene Aliases
H2-ALPHA
UniGene
Hs.503749
Gene Chromosome Location
Chr.2: 131476007-131482934 on Build GRCh38
Transcripts
XM_005263742.2 NM_080386.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3566529 c.64G>A p.E22K Chr.2: 131478224-131478224 on Build GRCh38
6006781 c.5G>T p.R2L Chr.2: 131478165-131478165 on Build GRCh38
3797972 c.80A>G p.E27G Chr.2: 131478240-131478240 on Build GRCh38
6813982 c.108G>A p.M36I Chr.2: 131478268-131478268 on Build GRCh38
3566530 c.113G>A p.S38N Chr.2: 131478273-131478273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772684160 Chr.2: 131478257-131478257 on Build GRCh38
rs547963721 Chr.2: 131478127-131478127 on Build GRCh38
rs757420147 Chr.2: 131478217-131478217 on Build GRCh38
rs2695519 Chr.2: 131478155-131478155 on Build GRCh38
rs759581201 Chr.2: 131478199-131478199 on Build GRCh38
rs761559557 Chr.2: 131478119-131478119 on Build GRCh38
rs762631194 Chr.2: 131478189-131478189 on Build GRCh38
rs760926055 Chr.2: 131478180-131478180 on Build GRCh38
rs569650706 Chr.2: 131478169-131478169 on Build GRCh38
rs762539966 Chr.2: 131478135-131478135 on Build GRCh38
rs2621996 Chr.2: 131478144-131478144 on Build GRCh38
rs905314694 Chr.2: 131478256-131478256 on Build GRCh38
rs778904212 Chr.2: 131478166-131478166 on Build GRCh38
rs772996766 Chr.2: 131478185-131478185 on Build GRCh38
rs146767427 Chr.2: 131478176-131478176 on Build GRCh38
rs764765776 Chr.2: 131478122-131478122 on Build GRCh38
rs754909176 Chr.2: 131478147-131478147 on Build GRCh38
rs772419712 Chr.2: 131478195-131478195 on Build GRCh38
rs778995974 Chr.2: 131478243-131478243 on Build GRCh38
rs771001531 Chr.2: 131478165-131478165 on Build GRCh38
rs745916475 Chr.2: 131478255-131478255 on Build GRCh38
rs377460015 Chr.2: 131478164-131478164 on Build GRCh38
rs753605346 Chr.2: 131478256-131478258 on Build GRCh38
rs766494367 Chr.2: 131478136-131478136 on Build GRCh38
rs530707778 Chr.2: 131478209-131478209 on Build GRCh38
rs781067007 Chr.2: 131478148-131478148 on Build GRCh38
rs149176469 Chr.2: 131478167-131478167 on Build GRCh38
rs201459956 Chr.2: 131478100-131478100 on Build GRCh38
rs756404051 Chr.2: 131478158-131478158 on Build GRCh38
rs769550209 Chr.2: 131478184-131478184 on Build GRCh38
rs777842519 Chr.2: 131478162-131478162 on Build GRCh38
rs986415142 Chr.2: 131478097-131478097 on Build GRCh38
rs912333410 Chr.2: 131478131-131478131 on Build GRCh38
rs780414369 Chr.2: 131478259-131478259 on Build GRCh38
rs147118488 Chr.2: 131478213-131478213 on Build GRCh38
rs754071611 Chr.2: 131478215-131478215 on Build GRCh38
rs767332287 Chr.2: 131478208-131478208 on Build GRCh38
rs931240112 Chr.2: 131478120-131478120 on Build GRCh38
rs751068564 Chr.2: 131478196-131478196 on Build GRCh38
rs2695520 Chr.2: 131478072-131478072 on Build GRCh38

View Full Product Details