Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42765433-42765668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSG8
Entrez Gene ID
440533
Gene Name
pregnancy specific beta-1-glycoprotein 8
Gene Aliases
PSG1
UniGene
Hs.466843
Gene Chromosome Location
Chr.19: 42752687-42765692 on Build GRCh38
Transcripts
NM_001130167.1 NM_001130168.1 NM_182707.2 XM_011526986.1 XM_011526988.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3534952 c.22C>T p.P8S Chr.19: 42765560-42765560 on Build GRCh38
3534950 c.45G>A p.W15* Chr.19: 42765537-42765537 on Build GRCh38
121662 c.54C>G p.L18L Chr.19: 42765528-42765528 on Build GRCh38
6292015 c.8T>C p.L3P Chr.19: 42765574-42765574 on Build GRCh38
3404309 c.64G>T p.A22S Chr.19: 42765518-42765518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779444955 Chr.19: 42765580-42765580 on Build GRCh38
rs765376973 Chr.19: 42765507-42765507 on Build GRCh38
rs751977560 Chr.19: 42765481-42765481 on Build GRCh38
rs749522143 Chr.19: 42765607-42765607 on Build GRCh38
rs375141281 Chr.19: 42765497-42765497 on Build GRCh38
rs780499586 Chr.19: 42765478-42765478 on Build GRCh38
rs757986183 Chr.19: 42765575-42765575 on Build GRCh38
rs746863859 Chr.19: 42765502-42765502 on Build GRCh38
rs575528307 Chr.19: 42765495-42765495 on Build GRCh38
rs148711271 Chr.19: 42765469-42765469 on Build GRCh38
rs34129574 Chr.19: 42765552-42765553 on Build GRCh38
rs142949326 Chr.19: 42765541-42765541 on Build GRCh38
rs775265999 Chr.19: 42765500-42765500 on Build GRCh38
rs375994401 Chr.19: 42765605-42765605 on Build GRCh38
rs145793435 Chr.19: 42765530-42765530 on Build GRCh38
rs756998284 Chr.19: 42765532-42765532 on Build GRCh38
rs7246055 Chr.19: 42765486-42765486 on Build GRCh38
rs755265154 Chr.19: 42765484-42765484 on Build GRCh38
rs752003225 Chr.19: 42765525-42765525 on Build GRCh38
rs771114706 Chr.19: 42765610-42765610 on Build GRCh38
rs746461659 Chr.19: 42765590-42765590 on Build GRCh38
rs778566389 Chr.19: 42765493-42765493 on Build GRCh38
rs754197910 Chr.19: 42765627-42765627 on Build GRCh38
rs554451127 Chr.19: 42765592-42765592 on Build GRCh38
rs373909679 Chr.19: 42765574-42765574 on Build GRCh38
rs987012529 Chr.19: 42765644-42765644 on Build GRCh38
rs781138692 Chr.19: 42765593-42765593 on Build GRCh38
rs760336269 Chr.19: 42765467-42765467 on Build GRCh38
rs753133021 Chr.19: 42765573-42765573 on Build GRCh38
rs538084246 Chr.19: 42765621-42765621 on Build GRCh38
rs768334685 Chr.19: 42765503-42765503 on Build GRCh38
rs369010869 Chr.19: 42765616-42765616 on Build GRCh38
rs757952650 Chr.19: 42765540-42765540 on Build GRCh38
rs368091087 Chr.19: 42765471-42765471 on Build GRCh38
rs61393109 Chr.19: 42765534-42765534 on Build GRCh38
rs768581313 Chr.19: 42765546-42765546 on Build GRCh38
rs918823242 Chr.19: 42765483-42765483 on Build GRCh38
rs773403623 Chr.19: 42765549-42765549 on Build GRCh38
rs71337226 Chr.19: 42765547-42765553 on Build GRCh38
rs747988704 Chr.19: 42765594-42765594 on Build GRCh38
rs776973571 Chr.19: 42765504-42765504 on Build GRCh38
rs753979788 Chr.19: 42765468-42765468 on Build GRCh38
rs1010143443 Chr.19: 42765535-42765535 on Build GRCh38
rs762973584 Chr.19: 42765517-42765517 on Build GRCh38
rs763084223 Chr.19: 42765551-42765551 on Build GRCh38
rs748878349 Chr.19: 42765487-42765487 on Build GRCh38
rs770649449 Chr.19: 42765492-42765492 on Build GRCh38
rs7245423 Chr.19: 42765553-42765553 on Build GRCh38
rs569073584 Chr.19: 42765620-42765620 on Build GRCh38
rs759610460 Chr.19: 42765614-42765614 on Build GRCh38
rs536412761 Chr.19: 42765585-42765585 on Build GRCh38
rs759960457 Chr.19: 42765556-42765556 on Build GRCh38
rs369084125 Chr.19: 42765528-42765528 on Build GRCh38
rs7260508 Chr.19: 42765547-42765547 on Build GRCh38
rs774326303 Chr.19: 42765612-42765612 on Build GRCh38
rs755427646 Chr.19: 42765526-42765526 on Build GRCh38
rs146791116 Chr.19: 42765559-42765559 on Build GRCh38
rs762039774 Chr.19: 42765505-42765505 on Build GRCh38
rs758912009 Chr.19: 42765475-42765475 on Build GRCh38
rs762517047 Chr.19: 42765628-42765628 on Build GRCh38
rs200337862 Chr.19: 42765597-42765597 on Build GRCh38
rs772403302 Chr.19: 42765595-42765596 on Build GRCh38
rs776107045 Chr.19: 42765618-42765618 on Build GRCh38
rs551635334 Chr.19: 42765641-42765641 on Build GRCh38
rs773313488 Chr.19: 42765514-42765514 on Build GRCh38
rs200221981 Chr.19: 42765548-42765548 on Build GRCh38
rs62112127 Chr.19: 42765552-42765552 on Build GRCh38
rs748571020 Chr.19: 42765590-42765593 on Build GRCh38
rs963052145 Chr.19: 42765582-42765582 on Build GRCh38

View Full Product Details