Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3005912-3006137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLE2
Entrez Gene ID
7089
Gene Name
transducin like enhancer of split 2
Gene Aliases
ESG, ESG2, GRG2
UniGene
Hs.332173
Gene Chromosome Location
Chr.19: 2997638-3047635 on Build GRCh38
Transcripts
NM_001144761.1 NM_001144762.1 NM_001300846.1 NM_003260.4 XM_006722864.2 XM_011528230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3959874 c.1529A>C p.K510T Chr.19: 3005940-3005940 on Build GRCh38
5855359 c.1501-1G>A p.? Chr.19: 3005969-3005969 on Build GRCh38
6802202 c.1505G>A p.R502Q Chr.19: 3005964-3005964 on Build GRCh38
4979954 c.1546C>T p.R516W Chr.19: 3005923-3005923 on Build GRCh38
4076561 c.1544G>A p.G515D Chr.19: 3005925-3005925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771825948 Chr.19: 3005976-3005976 on Build GRCh38
rs779804672 Chr.19: 3006088-3006088 on Build GRCh38
rs562202032 Chr.19: 3006112-3006112 on Build GRCh38
rs750145078 Chr.19: 3006057-3006062 on Build GRCh38
rs369289668 Chr.19: 3006058-3006058 on Build GRCh38
rs780906862 Chr.19: 3006100-3006100 on Build GRCh38
rs375716302 Chr.19: 3006095-3006095 on Build GRCh38
rs755960327 Chr.19: 3006085-3006107 on Build GRCh38
rs921154962 Chr.19: 3006038-3006038 on Build GRCh38
rs773032281 Chr.19: 3006107-3006107 on Build GRCh38
rs779857584 Chr.19: 3005977-3005977 on Build GRCh38
rs373360448 Chr.19: 3005931-3005931 on Build GRCh38
rs774528805 Chr.19: 3006117-3006117 on Build GRCh38
rs754164250 Chr.19: 3006086-3006086 on Build GRCh38
rs772666995 Chr.19: 3005962-3005962 on Build GRCh38
rs759574713 Chr.19: 3006063-3006063 on Build GRCh38
rs932047852 Chr.19: 3006019-3006019 on Build GRCh38
rs377534888 Chr.19: 3005953-3005953 on Build GRCh38
rs756705307 Chr.19: 3006078-3006078 on Build GRCh38
rs754513347 Chr.19: 3006096-3006096 on Build GRCh38
rs962570022 Chr.19: 3005965-3005965 on Build GRCh38
rs1043480911 Chr.19: 3006083-3006083 on Build GRCh38
rs774575811 Chr.19: 3006059-3006059 on Build GRCh38
rs550449761 Chr.19: 3006082-3006082 on Build GRCh38
rs577308167 Chr.19: 3006106-3006106 on Build GRCh38
rs752135907 Chr.19: 3006114-3006114 on Build GRCh38
rs762657075 Chr.19: 3006048-3006048 on Build GRCh38
rs781625359 Chr.19: 3005954-3005954 on Build GRCh38
rs375003469 Chr.19: 3005974-3005974 on Build GRCh38
rs200315276 Chr.19: 3005944-3005944 on Build GRCh38
rs1005331740 Chr.19: 3006061-3006061 on Build GRCh38
rs767627130 Chr.19: 3006066-3006066 on Build GRCh38
rs769408769 Chr.19: 3005930-3005930 on Build GRCh38
rs768305002 Chr.19: 3006010-3006010 on Build GRCh38
rs530394864 Chr.19: 3005966-3005966 on Build GRCh38
rs368808722 Chr.19: 3006113-3006113 on Build GRCh38
rs773122564 Chr.19: 3006044-3006044 on Build GRCh38
rs755062143 Chr.19: 3005951-3005951 on Build GRCh38
rs770741631 Chr.19: 3006114-3006114 on Build GRCh38
rs769462624 Chr.19: 3006043-3006043 on Build GRCh38
rs778253232 Chr.19: 3005972-3005972 on Build GRCh38
rs925477261 Chr.19: 3006093-3006093 on Build GRCh38
rs776676299 Chr.19: 3006011-3006011 on Build GRCh38
rs761858295 Chr.19: 3006015-3006015 on Build GRCh38
rs372734700 Chr.19: 3006072-3006072 on Build GRCh38
rs746462595 Chr.19: 3005992-3005992 on Build GRCh38

View Full Product Details