Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113801252-113801519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP28
Entrez Gene ID
57646
Gene Name
ubiquitin specific peptidase 28
Gene Aliases
-
UniGene
Hs.503891
Gene Chromosome Location
Chr.11: 113797875-113875580 on Build GRCh38
Transcripts
NM_001301029.1 NM_020886.3 XM_005271630.2 XM_005271631.2 XM_005271632.2 XM_005271633.4 XM_005271636.2 XM_005271637.2 XM_005271638.2 XM_005271639.2 XM_011542936.2 XM_011542938.1 XM_011542941.1 XM_011542942.1 XM_017018056.1 XM_017018057.1 XM_017018058.1 XM_017018059.1 XM_017018060.1 XM_017018061.1 XM_017018062.1 XM_017018063.1 XM_017018064.1 XM_017018065.1 XM_017018066.1 XM_017018067.1 XM_017018068.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923233 c.3033C>A p.C1011* Chr.11: 113801508-113801508 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs915788430 Chr.11: 113801390-113801390 on Build GRCh38
rs527868617 Chr.11: 113801387-113801387 on Build GRCh38
rs564855273 Chr.11: 113801280-113801280 on Build GRCh38
rs201322816 Chr.11: 113801495-113801495 on Build GRCh38
rs902362193 Chr.11: 113801294-113801294 on Build GRCh38
rs144977731 Chr.11: 113801497-113801497 on Build GRCh38
rs761759408 Chr.11: 113801358-113801358 on Build GRCh38
rs1016938173 Chr.11: 113801422-113801422 on Build GRCh38
rs751914539 Chr.11: 113801440-113801440 on Build GRCh38
rs750361252 Chr.11: 113801493-113801493 on Build GRCh38
rs763797981 Chr.11: 113801461-113801461 on Build GRCh38
rs898549495 Chr.11: 113801496-113801496 on Build GRCh38
rs200554152 Chr.11: 113801477-113801477 on Build GRCh38
rs908670771 Chr.11: 113801433-113801433 on Build GRCh38
rs755831902 Chr.11: 113801449-113801449 on Build GRCh38
rs1056432410 Chr.11: 113801389-113801389 on Build GRCh38
rs557521524 Chr.11: 113801331-113801332 on Build GRCh38
rs767257277 Chr.11: 113801421-113801421 on Build GRCh38
rs1042223183 Chr.11: 113801281-113801281 on Build GRCh38
rs895137488 Chr.11: 113801411-113801411 on Build GRCh38
rs756762753 Chr.11: 113801486-113801486 on Build GRCh38
rs939230097 Chr.11: 113801301-113801301 on Build GRCh38
rs547712865 Chr.11: 113801413-113801413 on Build GRCh38
rs940079721 Chr.11: 113801476-113801476 on Build GRCh38
rs778784065 Chr.11: 113801491-113801491 on Build GRCh38

View Full Product Details