Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34095523-34095782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS6
Entrez Gene ID
64968
Gene Name
mitochondrial ribosomal protein S6
Gene Aliases
C21orf101, MRP-S6, RPMS6, S6mt
UniGene
Hs.302742
Gene Chromosome Location
Chr.21: 34073523-34143034 on Build GRCh38
Transcripts
NM_032476.3 NM_006933.6

Gene Symbol
SLC5A3
Entrez Gene ID
6526
Gene Name
solute carrier family 5 member 3
Gene Aliases
BCW2, SMIT, SMIT1, SMIT2
UniGene
Hs.302742
Gene Chromosome Location
Chr.21: 34073523-34106262 on Build GRCh38
Transcripts
NM_032476.3 NM_006933.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4494839 c.440C>T p.A147V Chr.21: 34095638-34095638 on Build GRCh38
5558616 c.443T>C p.L148P Chr.21: 34095641-34095641 on Build GRCh38
5558508 c.387G>A p.L129L Chr.21: 34095585-34095585 on Build GRCh38
6783103 c.443delT p.F149fs*20 Chr.21: 34095641-34095641 on Build GRCh38
4501054 c.579C>T p.I193I Chr.21: 34095777-34095777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764401129 Chr.21: 34095639-34095639 on Build GRCh38
rs370993290 Chr.21: 34095581-34095581 on Build GRCh38
rs754822342 Chr.21: 34095753-34095753 on Build GRCh38
rs150670040 Chr.21: 34095681-34095681 on Build GRCh38
rs367913360 Chr.21: 34095723-34095723 on Build GRCh38
rs541107774 Chr.21: 34095615-34095615 on Build GRCh38
rs747641336 Chr.21: 34095572-34095572 on Build GRCh38
rs143944881 Chr.21: 34095625-34095625 on Build GRCh38
rs961234819 Chr.21: 34095552-34095552 on Build GRCh38
rs746392853 Chr.21: 34095618-34095618 on Build GRCh38
rs371781200 Chr.21: 34095747-34095747 on Build GRCh38
rs201799945 Chr.21: 34095741-34095741 on Build GRCh38
rs368643708 Chr.21: 34095558-34095558 on Build GRCh38
rs780763091 Chr.21: 34095755-34095755 on Build GRCh38
rs761169491 Chr.21: 34095637-34095637 on Build GRCh38
rs750330466 Chr.21: 34095702-34095702 on Build GRCh38
rs766375586 Chr.21: 34095729-34095729 on Build GRCh38
rs760198872 Chr.21: 34095588-34095588 on Build GRCh38
rs374318642 Chr.21: 34095633-34095633 on Build GRCh38
rs752349480 Chr.21: 34095759-34095759 on Build GRCh38
rs1027342132 Chr.21: 34095599-34095599 on Build GRCh38
rs761920535 Chr.21: 34095682-34095682 on Build GRCh38
rs765443139 Chr.21: 34095688-34095688 on Build GRCh38

View Full Product Details