Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15213889-15214128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM171A1
Entrez Gene ID
221061
Gene Name
family with sequence similarity 171 member A1
Gene Aliases
C10orf38
UniGene
Hs.66762
Gene Chromosome Location
Chr.10: 15211643-15374532 on Build GRCh38
Transcripts
NM_001010924.1 XM_011519378.2 XM_017015904.1 XM_017015905.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6129107 c.1625C>T p.S542L Chr.10: 15213963-15213963 on Build GRCh38
916697 c.1565C>T p.A522V Chr.10: 15214023-15214023 on Build GRCh38
1638499 c.1626G>A p.S542S Chr.10: 15213962-15213962 on Build GRCh38
427333 c.1530A>G p.G510G Chr.10: 15214058-15214058 on Build GRCh38
1627409 c.1473C>T p.T491T Chr.10: 15214115-15214115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796850715 Chr.10: 15213988-15213988 on Build GRCh38
rs891200971 Chr.10: 15213959-15213959 on Build GRCh38
rs3814166 Chr.10: 15213916-15213916 on Build GRCh38
rs747891159 Chr.10: 15213944-15213944 on Build GRCh38
rs769143641 Chr.10: 15214000-15214000 on Build GRCh38
rs776191844 Chr.10: 15213987-15213987 on Build GRCh38
rs752713972 Chr.10: 15214026-15214026 on Build GRCh38
rs769213278 Chr.10: 15214042-15214042 on Build GRCh38
rs147234872 Chr.10: 15213994-15213994 on Build GRCh38
rs368859404 Chr.10: 15214035-15214035 on Build GRCh38
rs947363709 Chr.10: 15214033-15214033 on Build GRCh38
rs866420092 Chr.10: 15214060-15214060 on Build GRCh38
rs760487071 Chr.10: 15214029-15214029 on Build GRCh38
rs777688592 Chr.10: 15214030-15214030 on Build GRCh38
rs771127014 Chr.10: 15214091-15214091 on Build GRCh38
rs1051211743 Chr.10: 15213943-15213943 on Build GRCh38
rs770538157 Chr.10: 15213975-15213975 on Build GRCh38
rs753920185 Chr.10: 15213932-15213932 on Build GRCh38
rs766573146 Chr.10: 15214023-15214023 on Build GRCh38
rs561182923 Chr.10: 15213935-15213935 on Build GRCh38
rs767672202 Chr.10: 15213922-15213922 on Build GRCh38
rs748630124 Chr.10: 15214070-15214070 on Build GRCh38
rs866877027 Chr.10: 15214072-15214072 on Build GRCh38
rs981899880 Chr.10: 15214008-15214008 on Build GRCh38
rs754938956 Chr.10: 15213933-15213933 on Build GRCh38
rs746630475 Chr.10: 15213972-15213972 on Build GRCh38
rs750671974 Chr.10: 15213925-15213925 on Build GRCh38
rs763843129 Chr.10: 15214062-15214062 on Build GRCh38
rs759547506 Chr.10: 15214024-15214024 on Build GRCh38
rs148721770 Chr.10: 15214036-15214036 on Build GRCh38
rs185961044 Chr.10: 15214080-15214080 on Build GRCh38
rs776002188 Chr.10: 15213952-15213954 on Build GRCh38
rs369876836 Chr.10: 15213929-15213929 on Build GRCh38
rs557564382 Chr.10: 15214079-15214079 on Build GRCh38
rs868521925 Chr.10: 15214039-15214039 on Build GRCh38
rs773613941 Chr.10: 15214022-15214022 on Build GRCh38
rs139974823 Chr.10: 15213917-15213917 on Build GRCh38
rs945430580 Chr.10: 15214017-15214017 on Build GRCh38
rs751370179 Chr.10: 15214031-15214031 on Build GRCh38
rs762119725 Chr.10: 15214010-15214010 on Build GRCh38
rs774698433 Chr.10: 15213918-15213918 on Build GRCh38
rs978903666 Chr.10: 15214044-15214044 on Build GRCh38
rs777528921 Chr.10: 15213963-15213963 on Build GRCh38
rs776738874 Chr.10: 15214105-15214105 on Build GRCh38
rs758289605 Chr.10: 15213960-15213960 on Build GRCh38
rs756328994 Chr.10: 15213928-15213928 on Build GRCh38
rs375846326 Chr.10: 15214021-15214021 on Build GRCh38
rs760950735 Chr.10: 15214084-15214084 on Build GRCh38
rs755359286 Chr.10: 15213972-15213973 on Build GRCh38
rs529830750 Chr.10: 15213962-15213962 on Build GRCh38
rs765216749 Chr.10: 15214025-15214025 on Build GRCh38
rs142443473 Chr.10: 15214074-15214074 on Build GRCh38
rs774855716 Chr.10: 15214003-15214003 on Build GRCh38
rs150161950 Chr.10: 15213921-15213921 on Build GRCh38

View Full Product Details