Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 8798038-8798299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9A
Entrez Gene ID
171482
Gene Name
family with sequence similarity 9 member A
Gene Aliases
TEX39A
UniGene
Hs.382062
Gene Chromosome Location
Chr.X: 8790795-8801383 on Build GRCh38
Transcripts
NM_001171186.1 NM_174951.3

SNP Detail

SNP ID Chromosome Location
rs768846920 Chr.X: 8798171-8798171 on Build GRCh38
rs748449822 Chr.X: 8798231-8798231 on Build GRCh38
rs770089129 Chr.X: 8798233-8798233 on Build GRCh38
rs372062687 Chr.X: 8798130-8798130 on Build GRCh38
rs745777995 Chr.X: 8798159-8798159 on Build GRCh38
rs757189875 Chr.X: 8798135-8798135 on Build GRCh38
rs913986395 Chr.X: 8798146-8798146 on Build GRCh38
rs183279511 Chr.X: 8798257-8798257 on Build GRCh38
rs755702503 Chr.X: 8798120-8798120 on Build GRCh38
rs755066530 Chr.X: 8798087-8798087 on Build GRCh38
rs761145686 Chr.X: 8798196-8798196 on Build GRCh38
rs765254049 Chr.X: 8798129-8798129 on Build GRCh38
rs777550388 Chr.X: 8798269-8798269 on Build GRCh38
rs186153255 Chr.X: 8798266-8798266 on Build GRCh38
rs926783631 Chr.X: 8798261-8798261 on Build GRCh38
rs2018496 Chr.X: 8798250-8798250 on Build GRCh38
rs949496496 Chr.X: 8798244-8798244 on Build GRCh38
rs368202802 Chr.X: 8798100-8798100 on Build GRCh38
rs778764949 Chr.X: 8798147-8798147 on Build GRCh38
rs773380253 Chr.X: 8798064-8798064 on Build GRCh38
rs747663083 Chr.X: 8798185-8798186 on Build GRCh38
rs776538849 Chr.X: 8798182-8798182 on Build GRCh38

View Full Product Details