Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39015408-39015647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3C
Entrez Gene ID
27350
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3C
Gene Aliases
A3C, APOBEC1L, ARDC2, ARDC4, ARP5, PBI, bK150C2.3
UniGene
Hs.441124
Gene Chromosome Location
Chr.22: 39014260-39018838 on Build GRCh38
Transcripts
NM_014508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6346752 c.36G>T p.M12I Chr.22: 39015613-39015613 on Build GRCh38
6602363 c.32C>T p.A11V Chr.22: 39015609-39015609 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs963595800 Chr.22: 39015507-39015507 on Build GRCh38
rs536763432 Chr.22: 39015462-39015462 on Build GRCh38
rs934658736 Chr.22: 39015517-39015517 on Build GRCh38
rs921505471 Chr.22: 39015495-39015495 on Build GRCh38
rs950448964 Chr.22: 39015455-39015455 on Build GRCh38
rs200291335 Chr.22: 39015555-39015555 on Build GRCh38
rs367840996 Chr.22: 39015600-39015600 on Build GRCh38
rs544167247 Chr.22: 39015601-39015601 on Build GRCh38
rs971142934 Chr.22: 39015434-39015434 on Build GRCh38
rs747545111 Chr.22: 39015579-39015579 on Build GRCh38
rs1054541817 Chr.22: 39015531-39015531 on Build GRCh38
rs1025956153 Chr.22: 39015450-39015450 on Build GRCh38
rs760801793 Chr.22: 39015559-39015559 on Build GRCh38
rs973638896 Chr.22: 39015538-39015538 on Build GRCh38
rs577991980 Chr.22: 39015431-39015431 on Build GRCh38
rs575285769 Chr.22: 39015512-39015512 on Build GRCh38
rs201273021 Chr.22: 39015550-39015550 on Build GRCh38
rs868429335 Chr.22: 39015484-39015484 on Build GRCh38
rs752706550 Chr.22: 39015558-39015558 on Build GRCh38
rs764276625 Chr.22: 39015560-39015560 on Build GRCh38
rs984523656 Chr.22: 39015467-39015467 on Build GRCh38
rs774083173 Chr.22: 39015553-39015553 on Build GRCh38
rs769224850 Chr.22: 39015583-39015583 on Build GRCh38
rs762553668 Chr.22: 39015556-39015556 on Build GRCh38
rs754575706 Chr.22: 39015555-39015556 on Build GRCh38
rs370883473 Chr.22: 39015545-39015545 on Build GRCh38
rs758913511 Chr.22: 39015572-39015572 on Build GRCh38
rs775228848 Chr.22: 39015619-39015619 on Build GRCh38
rs563276446 Chr.22: 39015496-39015496 on Build GRCh38
rs981054523 Chr.22: 39015435-39015435 on Build GRCh38
rs750789262 Chr.22: 39015569-39015569 on Build GRCh38
rs908959809 Chr.22: 39015472-39015472 on Build GRCh38
rs375901753 Chr.22: 39015568-39015568 on Build GRCh38
rs780375676 Chr.22: 39015577-39015577 on Build GRCh38
rs774174614 Chr.22: 39015611-39015611 on Build GRCh38
rs779062792 Chr.22: 39015487-39015487 on Build GRCh38
rs926881673 Chr.22: 39015500-39015500 on Build GRCh38
rs771925073 Chr.22: 39015616-39015616 on Build GRCh38
rs943046085 Chr.22: 39015567-39015567 on Build GRCh38
rs757607314 Chr.22: 39015562-39015562 on Build GRCh38
rs145728799 Chr.22: 39015614-39015614 on Build GRCh38
rs371604220 Chr.22: 39015615-39015615 on Build GRCh38
rs376666585 Chr.22: 39015551-39015551 on Build GRCh38

View Full Product Details