Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39500522-39500773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL3
Entrez Gene ID
10683
Gene Name
delta like canonical Notch ligand 3
Gene Aliases
SCDO1
UniGene
Hs.127792
Gene Chromosome Location
Chr.19: 39498917-39508481 on Build GRCh38
Transcripts
NM_016941.3 NM_203486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5930363 c.383G>T p.R128I Chr.19: 39500646-39500646 on Build GRCh38
4546791 c.407G>A p.G136E Chr.19: 39500670-39500670 on Build GRCh38
3378736 c.387G>T p.E129D Chr.19: 39500650-39500650 on Build GRCh38
3533797 c.373G>A p.E125K Chr.19: 39500636-39500636 on Build GRCh38
4608752 c.406G>T p.G136* Chr.19: 39500669-39500669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2304223 Chr.19: 39500597-39500597 on Build GRCh38
rs770966935 Chr.19: 39500636-39500636 on Build GRCh38
rs377405105 Chr.19: 39500733-39500733 on Build GRCh38
rs975719833 Chr.19: 39500553-39500553 on Build GRCh38
rs761188289 Chr.19: 39500593-39500593 on Build GRCh38
rs372367758 Chr.19: 39500723-39500723 on Build GRCh38
rs186443050 Chr.19: 39500717-39500717 on Build GRCh38
rs767029113 Chr.19: 39500595-39500595 on Build GRCh38
rs761454301 Chr.19: 39500657-39500657 on Build GRCh38
rs201180653 Chr.19: 39500722-39500722 on Build GRCh38
rs775870175 Chr.19: 39500656-39500658 on Build GRCh38
rs765205227 Chr.19: 39500642-39500642 on Build GRCh38
rs760054689 Chr.19: 39500576-39500582 on Build GRCh38
rs777813964 Chr.19: 39500633-39500633 on Build GRCh38
rs201227250 Chr.19: 39500691-39500691 on Build GRCh38
rs754371527 Chr.19: 39500596-39500596 on Build GRCh38
rs572411769 Chr.19: 39500745-39500745 on Build GRCh38
rs745793784 Chr.19: 39500672-39500672 on Build GRCh38
rs2304222 Chr.19: 39500563-39500563 on Build GRCh38
rs762606145 Chr.19: 39500686-39500686 on Build GRCh38
rs201902809 Chr.19: 39500600-39500600 on Build GRCh38
rs763594072 Chr.19: 39500573-39500573 on Build GRCh38
rs758663865 Chr.19: 39500615-39500615 on Build GRCh38
rs770120266 Chr.19: 39500621-39500621 on Build GRCh38
rs769650774 Chr.19: 39500673-39500673 on Build GRCh38
rs368117513 Chr.19: 39500708-39500708 on Build GRCh38
rs773840011 Chr.19: 39500700-39500700 on Build GRCh38
rs760127403 Chr.19: 39500716-39500716 on Build GRCh38
rs775367877 Chr.19: 39500679-39500679 on Build GRCh38
rs754461205 Chr.19: 39500709-39500709 on Build GRCh38
rs761431149 Chr.19: 39500704-39500704 on Build GRCh38
rs995675341 Chr.19: 39500678-39500678 on Build GRCh38
rs143468961 Chr.19: 39500635-39500635 on Build GRCh38
rs751102073 Chr.19: 39500589-39500589 on Build GRCh38
rs928532876 Chr.19: 39500599-39500599 on Build GRCh38
rs913536356 Chr.19: 39500591-39500591 on Build GRCh38
rs113780398 Chr.19: 39500653-39500653 on Build GRCh38

View Full Product Details