Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22250914-22251181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2K
Entrez Gene ID
29904
Gene Name
eukaryotic elongation factor 2 kinase
Gene Aliases
HSU93850, eEF-2K
UniGene
Hs.498892
Gene Chromosome Location
Chr.16: 22206264-22288745 on Build GRCh38
Transcripts
NM_013302.3 XM_017023193.1 XM_017023194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1629824 c.473C>T p.A158V Chr.16: 22251177-22251177 on Build GRCh38
3507632 c.454C>T p.L152F Chr.16: 22251158-22251158 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs140219296 Chr.16: 22251019-22251019 on Build GRCh38
rs746018935 Chr.16: 22251153-22251153 on Build GRCh38
rs562798620 Chr.16: 22251098-22251098 on Build GRCh38
rs375705424 Chr.16: 22251086-22251086 on Build GRCh38
rs752609933 Chr.16: 22251132-22251132 on Build GRCh38
rs760516944 Chr.16: 22251121-22251122 on Build GRCh38
rs777550326 Chr.16: 22251149-22251149 on Build GRCh38
rs766201074 Chr.16: 22251122-22251122 on Build GRCh38
rs371491303 Chr.16: 22251131-22251131 on Build GRCh38
rs1049968414 Chr.16: 22250941-22250941 on Build GRCh38
rs751189128 Chr.16: 22251128-22251128 on Build GRCh38
rs751136394 Chr.16: 22251147-22251147 on Build GRCh38
rs765438321 Chr.16: 22251134-22251134 on Build GRCh38
rs961177726 Chr.16: 22251130-22251130 on Build GRCh38
rs543207264 Chr.16: 22251089-22251089 on Build GRCh38
rs369010030 Chr.16: 22251116-22251116 on Build GRCh38
rs763061605 Chr.16: 22250946-22250946 on Build GRCh38
rs904292176 Chr.16: 22251026-22251026 on Build GRCh38
rs764118978 Chr.16: 22250954-22250954 on Build GRCh38
rs1042725413 Chr.16: 22251022-22251022 on Build GRCh38
rs529460713 Chr.16: 22250993-22250993 on Build GRCh38
rs369413078 Chr.16: 22251148-22251148 on Build GRCh38
rs751804990 Chr.16: 22250977-22250977 on Build GRCh38
rs1026854446 Chr.16: 22251137-22251137 on Build GRCh38
rs998548918 Chr.16: 22251066-22251066 on Build GRCh38
rs772138145 Chr.16: 22251158-22251158 on Build GRCh38
rs1019684606 Chr.16: 22250982-22250982 on Build GRCh38

View Full Product Details