Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50290168-50290403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFRD2
Entrez Gene ID
7866
Gene Name
interferon related developmental regulator 2
Gene Aliases
IFNRP, SKMc15, SM15
UniGene
Hs.315177
Gene Chromosome Location
Chr.3: 50287732-50292595 on Build GRCh38
Transcripts
NM_006764.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6567336 c.868G>A p.E290K Chr.3: 50290188-50290188 on Build GRCh38
6567335 c.562G>A p.E188K Chr.3: 50290188-50290188 on Build GRCh38
6683967 c.490C>T p.R164C Chr.3: 50290260-50290260 on Build GRCh38
2851212 c.852G>T p.T284T Chr.3: 50290204-50290204 on Build GRCh38
1254627 c.814C>T p.R272C Chr.3: 50290242-50290242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199853119 Chr.3: 50290198-50290198 on Build GRCh38
rs782406511 Chr.3: 50290259-50290259 on Build GRCh38
rs781873482 Chr.3: 50290291-50290291 on Build GRCh38
rs782019385 Chr.3: 50290205-50290205 on Build GRCh38
rs782133623 Chr.3: 50290358-50290358 on Build GRCh38
rs782279131 Chr.3: 50290241-50290241 on Build GRCh38
rs199620289 Chr.3: 50290313-50290313 on Build GRCh38
rs367765510 Chr.3: 50290258-50290258 on Build GRCh38
rs782655489 Chr.3: 50290238-50290238 on Build GRCh38
rs782509554 Chr.3: 50290317-50290317 on Build GRCh38
rs782166320 Chr.3: 50290210-50290210 on Build GRCh38
rs148311115 Chr.3: 50290231-50290231 on Build GRCh38
rs782569493 Chr.3: 50290246-50290246 on Build GRCh38
rs781792600 Chr.3: 50290305-50290305 on Build GRCh38
rs781808384 Chr.3: 50290215-50290215 on Build GRCh38
rs782790588 Chr.3: 50290359-50290359 on Build GRCh38
rs782048206 Chr.3: 50290366-50290366 on Build GRCh38
rs369427457 Chr.3: 50290330-50290330 on Build GRCh38
rs782816152 Chr.3: 50290228-50290228 on Build GRCh38
rs371709943 Chr.3: 50290307-50290307 on Build GRCh38
rs782446572 Chr.3: 50290230-50290230 on Build GRCh38
rs377155857 Chr.3: 50290328-50290328 on Build GRCh38
rs782670972 Chr.3: 50290321-50290321 on Build GRCh38
rs782814182 Chr.3: 50290302-50290302 on Build GRCh38
rs201827780 Chr.3: 50290325-50290325 on Build GRCh38
rs1025780292 Chr.3: 50290264-50290264 on Build GRCh38
rs377504643 Chr.3: 50290380-50290380 on Build GRCh38
rs201133694 Chr.3: 50290343-50290343 on Build GRCh38
rs374859692 Chr.3: 50290256-50290256 on Build GRCh38
rs782601507 Chr.3: 50290260-50290260 on Build GRCh38
rs370188116 Chr.3: 50290324-50290324 on Build GRCh38
rs201440217 Chr.3: 50290234-50290234 on Build GRCh38
rs368320868 Chr.3: 50290242-50290242 on Build GRCh38
rs191789502 Chr.3: 50290371-50290371 on Build GRCh38
rs782336815 Chr.3: 50290190-50290190 on Build GRCh38
rs200829619 Chr.3: 50290254-50290254 on Build GRCh38
rs782217307 Chr.3: 50290247-50290247 on Build GRCh38
rs782472574 Chr.3: 50290306-50290306 on Build GRCh38
rs782673597 Chr.3: 50290237-50290237 on Build GRCh38
rs782708615 Chr.3: 50290283-50290283 on Build GRCh38
rs781833970 Chr.3: 50290232-50290232 on Build GRCh38
rs782548615 Chr.3: 50290301-50290301 on Build GRCh38
rs782605999 Chr.3: 50290197-50290197 on Build GRCh38
rs781955481 Chr.3: 50290212-50290212 on Build GRCh38
rs782502721 Chr.3: 50290379-50290379 on Build GRCh38
rs199899087 Chr.3: 50290211-50290211 on Build GRCh38
rs1055808579 Chr.3: 50290192-50290192 on Build GRCh38
rs782714501 Chr.3: 50290368-50290368 on Build GRCh38
rs587650948 Chr.3: 50290322-50290322 on Build GRCh38
rs782059087 Chr.3: 50290261-50290261 on Build GRCh38

View Full Product Details