Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 776831-777094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLOD4
Entrez Gene ID
51031
Gene Name
glyoxalase domain containing 4
Gene Aliases
C17orf25, CGI-150, HC71
UniGene
Hs.279061
Gene Chromosome Location
Chr.17: 759309-782331 on Build GRCh38
Transcripts
NM_016080.3 XM_006721540.2 XM_017024711.1 XM_017024712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5853344 c.306G>A p.M102I Chr.17: 776868-776868 on Build GRCh38
6253342 c.255C>A p.D85E Chr.17: 776874-776874 on Build GRCh38
6016359 c.261+9C>T p.? Chr.17: 776859-776859 on Build GRCh38
5699139 c.306+1G>T p.? Chr.17: 776867-776867 on Build GRCh38
5853343 c.261G>A p.M87I Chr.17: 776868-776868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745636525 Chr.17: 776861-776861 on Build GRCh38
rs116051522 Chr.17: 776880-776880 on Build GRCh38
rs528441946 Chr.17: 777043-777043 on Build GRCh38
rs774573072 Chr.17: 777012-777012 on Build GRCh38
rs115297676 Chr.17: 776899-776899 on Build GRCh38
rs146931627 Chr.17: 777021-777021 on Build GRCh38
rs377754311 Chr.17: 777014-777014 on Build GRCh38
rs768728303 Chr.17: 776878-776878 on Build GRCh38
rs753328981 Chr.17: 776910-776910 on Build GRCh38
rs115557783 Chr.17: 776871-776871 on Build GRCh38
rs747220485 Chr.17: 776874-776874 on Build GRCh38
rs747191303 Chr.17: 776954-776954 on Build GRCh38
rs141525142 Chr.17: 776946-776946 on Build GRCh38
rs775744051 Chr.17: 776866-776866 on Build GRCh38
rs745834587 Chr.17: 776927-776927 on Build GRCh38
rs771892537 Chr.17: 776863-776863 on Build GRCh38
rs748218284 Chr.17: 776982-776982 on Build GRCh38
rs571701603 Chr.17: 777065-777065 on Build GRCh38
rs778877234 Chr.17: 776859-776859 on Build GRCh38
rs114532494 Chr.17: 776898-776898 on Build GRCh38
rs952080689 Chr.17: 776913-776913 on Build GRCh38
rs371359904 Chr.17: 776960-776960 on Build GRCh38
rs143088969 Chr.17: 776889-776889 on Build GRCh38
rs1022689629 Chr.17: 776867-776867 on Build GRCh38
rs778787698 Chr.17: 776925-776925 on Build GRCh38
rs114535491 Chr.17: 776905-776905 on Build GRCh38
rs756805516 Chr.17: 776924-776924 on Build GRCh38
rs2273453 Chr.17: 777059-777059 on Build GRCh38
rs142209461 Chr.17: 776858-776858 on Build GRCh38
rs770035218 Chr.17: 776983-776983 on Build GRCh38
rs776319223 Chr.17: 776896-776896 on Build GRCh38
rs1035174176 Chr.17: 776890-776890 on Build GRCh38
rs141115825 Chr.17: 776908-776908 on Build GRCh38
rs73285725 Chr.17: 777072-777072 on Build GRCh38
rs140149712 Chr.17: 776900-776900 on Build GRCh38
rs114621791 Chr.17: 776976-776976 on Build GRCh38
rs771258218 Chr.17: 777007-777007 on Build GRCh38
rs752334770 Chr.17: 776901-776901 on Build GRCh38
rs758142345 Chr.17: 776937-776937 on Build GRCh38
rs753525289 Chr.17: 777026-777026 on Build GRCh38
rs761704810 Chr.17: 776948-776948 on Build GRCh38
rs768759871 Chr.17: 776959-776959 on Build GRCh38
rs905340262 Chr.17: 776872-776872 on Build GRCh38
rs763565492 Chr.17: 777003-777003 on Build GRCh38
rs867082183 Chr.17: 776949-776949 on Build GRCh38

View Full Product Details