Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136831828-136832055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABL6
Entrez Gene ID
55684
Gene Name
RAB, member RAS oncogene family-like 6
Gene Aliases
C9orf86, PARF, RBEL1, pp8875
UniGene
Hs.370555
Gene Chromosome Location
Chr.9: 136807922-136841187 on Build GRCh38
Transcripts
NM_001173988.1 NM_001173989.2 NM_024718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1106947 c.574C>T p.R192C Chr.9: 136831836-136831836 on Build GRCh38
1106948 c.808C>T p.R270C Chr.9: 136831836-136831836 on Build GRCh38
3413471 c.575G>A p.R192H Chr.9: 136831837-136831837 on Build GRCh38
3905843 c.574C>T p.R192C Chr.9: 136831836-136831836 on Build GRCh38
5098096 c.599+8C>T p.? Chr.9: 136831869-136831869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374025267 Chr.9: 136831898-136831898 on Build GRCh38
rs369523477 Chr.9: 136831888-136831888 on Build GRCh38
rs939184230 Chr.9: 136831919-136831919 on Build GRCh38
rs560851510 Chr.9: 136832022-136832022 on Build GRCh38
rs967842502 Chr.9: 136831991-136831991 on Build GRCh38
rs939284482 Chr.9: 136831912-136831912 on Build GRCh38
rs756040167 Chr.9: 136831870-136831871 on Build GRCh38
rs932695571 Chr.9: 136831954-136831955 on Build GRCh38
rs565086099 Chr.9: 136831971-136831971 on Build GRCh38
rs202104047 Chr.9: 136831907-136831907 on Build GRCh38
rs371430602 Chr.9: 136831847-136831847 on Build GRCh38
rs946722867 Chr.9: 136831930-136831930 on Build GRCh38
rs781297196 Chr.9: 136831908-136831908 on Build GRCh38
rs1044158553 Chr.9: 136831940-136831940 on Build GRCh38
rs777136840 Chr.9: 136831848-136831848 on Build GRCh38
rs373712822 Chr.9: 136831876-136831876 on Build GRCh38
rs980542860 Chr.9: 136831994-136831994 on Build GRCh38
rs773534214 Chr.9: 136831953-136831954 on Build GRCh38
rs1009261676 Chr.9: 136831987-136831987 on Build GRCh38
rs898603781 Chr.9: 136831948-136831948 on Build GRCh38
rs778588267 Chr.9: 136831893-136831893 on Build GRCh38
rs747927959 Chr.9: 136831854-136831854 on Build GRCh38
rs541124839 Chr.9: 136832016-136832016 on Build GRCh38
rs779600869 Chr.9: 136831846-136831846 on Build GRCh38
rs892115738 Chr.9: 136831986-136831986 on Build GRCh38
rs529995938 Chr.9: 136832029-136832029 on Build GRCh38
rs370994500 Chr.9: 136831926-136831926 on Build GRCh38
rs1022445456 Chr.9: 136831988-136831988 on Build GRCh38
rs1038493253 Chr.9: 136831859-136831859 on Build GRCh38
rs1008485314 Chr.9: 136831976-136831976 on Build GRCh38
rs753396717 Chr.9: 136831890-136831890 on Build GRCh38
rs908485419 Chr.9: 136832027-136832027 on Build GRCh38
rs546693704 Chr.9: 136832034-136832034 on Build GRCh38
rs774465778 Chr.9: 136831881-136831881 on Build GRCh38
rs771795020 Chr.9: 136831865-136831865 on Build GRCh38
rs544148112 Chr.9: 136832021-136832021 on Build GRCh38
rs750932123 Chr.9: 136831885-136831885 on Build GRCh38
rs569546887 Chr.9: 136831906-136831906 on Build GRCh38
rs746977093 Chr.9: 136831869-136831869 on Build GRCh38
rs758198701 Chr.9: 136831899-136831899 on Build GRCh38
rs572876438 Chr.9: 136831969-136831969 on Build GRCh38
rs749337064 Chr.9: 136831911-136831911 on Build GRCh38
rs145330836 Chr.9: 136831962-136831962 on Build GRCh38
rs369097592 Chr.9: 136831972-136831972 on Build GRCh38
rs147673388 Chr.9: 136831984-136831984 on Build GRCh38
rs778551130 Chr.9: 136831852-136831852 on Build GRCh38
rs375714213 Chr.9: 136831870-136831870 on Build GRCh38
rs891271972 Chr.9: 136831973-136831973 on Build GRCh38
rs5901109 Chr.9: 136832014-136832014 on Build GRCh38
rs765847365 Chr.9: 136831889-136831889 on Build GRCh38
rs377675795 Chr.9: 136831884-136831884 on Build GRCh38
rs777708293 Chr.9: 136831868-136831868 on Build GRCh38
rs754508678 Chr.9: 136831891-136831891 on Build GRCh38
rs375261983 Chr.9: 136831873-136831873 on Build GRCh38
rs992085444 Chr.9: 136831913-136831913 on Build GRCh38
rs940071547 Chr.9: 136831922-136831922 on Build GRCh38
rs80220337 Chr.9: 136831954-136831954 on Build GRCh38
rs763243314 Chr.9: 136831875-136831875 on Build GRCh38
rs939998568 Chr.9: 136832033-136832033 on Build GRCh38
rs139987410 Chr.9: 136831914-136831914 on Build GRCh38
rs531056038 Chr.9: 136831997-136831997 on Build GRCh38
rs1004351459 Chr.9: 136831983-136831984 on Build GRCh38

View Full Product Details