Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113894904-113895177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4B1
Entrez Gene ID
10717
Gene Name
adaptor related protein complex 4 beta 1 subunit
Gene Aliases
BETA-4, CPSQ5, SPG47
UniGene
Hs.515048
Gene Chromosome Location
Chr.1: 113894194-113905202 on Build GRCh38
Transcripts
NM_001253852.2 NM_001253853.2 NM_001308312.1 NM_006594.4 XM_011540523.2 XM_011540524.2 XM_011540525.2 XM_011540528.2 XM_017000088.1 XM_017000089.1 XM_017000090.1 XM_017000091.1 XM_017000092.1 NR_037864.1 NR_125965.1

Gene Symbol
AP4B1-AS1
Entrez Gene ID
100287722
Gene Name
AP4B1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.664669
Gene Chromosome Location
Chr.1: 113812612-113901237 on Build GRCh38
Transcripts
NM_001253852.2 NM_001253853.2 NM_001308312.1 NM_006594.4 XM_011540523.2 XM_011540524.2 XM_011540525.2 XM_011540528.2 XM_017000088.1 XM_017000089.1 XM_017000090.1 XM_017000091.1 XM_017000092.1 NR_037864.1 NR_125965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5612240 c.2116A>G p.M706V Chr.1: 113895169-113895169 on Build GRCh38
1332826 c.2156C>T p.T719M Chr.1: 113895129-113895129 on Build GRCh38
1183147 c.2197A>G p.T733A Chr.1: 113895088-113895088 on Build GRCh38
3740544 c.2111C>T p.S704L Chr.1: 113895174-113895174 on Build GRCh38
5377235 c.2195G>A p.G732E Chr.1: 113895090-113895090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770654389 Chr.1: 113895024-113895024 on Build GRCh38
rs775133436 Chr.1: 113895046-113895046 on Build GRCh38
rs145170032 Chr.1: 113895134-113895134 on Build GRCh38
rs79050956 Chr.1: 113895085-113895085 on Build GRCh38
rs777639673 Chr.1: 113895123-113895123 on Build GRCh38
rs1048645653 Chr.1: 113894980-113894980 on Build GRCh38
rs759087184 Chr.1: 113895032-113895032 on Build GRCh38
rs888265544 Chr.1: 113895013-113895016 on Build GRCh38
rs564093511 Chr.1: 113895129-113895129 on Build GRCh38
rs760708371 Chr.1: 113895143-113895143 on Build GRCh38
rs899106341 Chr.1: 113894944-113894944 on Build GRCh38
rs370272880 Chr.1: 113895003-113895003 on Build GRCh38
rs769542224 Chr.1: 113895034-113895034 on Build GRCh38
rs752709086 Chr.1: 113895099-113895099 on Build GRCh38
rs199879996 Chr.1: 113895135-113895135 on Build GRCh38
rs752550982 Chr.1: 113895053-113895053 on Build GRCh38
rs754282318 Chr.1: 113895079-113895079 on Build GRCh38
rs11349 Chr.1: 113895115-113895115 on Build GRCh38
rs372937961 Chr.1: 113895074-113895074 on Build GRCh38
rs750985228 Chr.1: 113895057-113895057 on Build GRCh38
rs969793953 Chr.1: 113895140-113895140 on Build GRCh38
rs373927491 Chr.1: 113894959-113894959 on Build GRCh38
rs558088187 Chr.1: 113895054-113895054 on Build GRCh38
rs543728602 Chr.1: 113895097-113895097 on Build GRCh38
rs993347556 Chr.1: 113894963-113894963 on Build GRCh38
rs773137329 Chr.1: 113895020-113895020 on Build GRCh38
rs192845390 Chr.1: 113895048-113895048 on Build GRCh38
rs776530752 Chr.1: 113895031-113895031 on Build GRCh38
rs776274646 Chr.1: 113894962-113894962 on Build GRCh38
rs1038295064 Chr.1: 113894943-113894943 on Build GRCh38
rs761056707 Chr.1: 113895058-113895058 on Build GRCh38
rs148223090 Chr.1: 113895121-113895121 on Build GRCh38
rs746759703 Chr.1: 113895022-113895022 on Build GRCh38
rs750073101 Chr.1: 113895061-113895061 on Build GRCh38
rs993907837 Chr.1: 113894967-113894967 on Build GRCh38

View Full Product Details