Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47376915-47377182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPCAM
Entrez Gene ID
4072
Gene Name
epithelial cell adhesion molecule
Gene Aliases
DIAR5, EGP-2, EGP314, EGP40, ESA, HNPCC8, KS1/4, KSA, M4S1, MIC18, MK-1, TACSTD1, TROP1
UniGene
Hs.542050
Gene Chromosome Location
Chr.2: 47369148-47387028 on Build GRCh38
Transcripts
NM_002354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4550602 c.504G>A p.K168K Chr.2: 47377026-47377026 on Build GRCh38
148851 c.515C>T p.T172M Chr.2: 47377037-47377037 on Build GRCh38
1021237 c.521A>G p.Y174C Chr.2: 47377043-47377043 on Build GRCh38
5580958 c.510C>T p.I170I Chr.2: 47377032-47377032 on Build GRCh38
1021238 c.552T>A p.I184I Chr.2: 47377074-47377074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371372017 Chr.2: 47377038-47377038 on Build GRCh38
rs760391956 Chr.2: 47377121-47377121 on Build GRCh38
rs74531854 Chr.2: 47377037-47377037 on Build GRCh38
rs779059677 Chr.2: 47377029-47377029 on Build GRCh38
rs771569331 Chr.2: 47377040-47377040 on Build GRCh38
rs748471427 Chr.2: 47376971-47376971 on Build GRCh38
rs761508741 Chr.2: 47377105-47377105 on Build GRCh38
rs766261189 Chr.2: 47377016-47377016 on Build GRCh38
rs1028625705 Chr.2: 47376962-47376962 on Build GRCh38
rs373453367 Chr.2: 47377091-47377091 on Build GRCh38
rs1000898513 Chr.2: 47376947-47376947 on Build GRCh38
rs953208357 Chr.2: 47377059-47377059 on Build GRCh38
rs747286107 Chr.2: 47377039-47377039 on Build GRCh38
rs370932088 Chr.2: 47377112-47377112 on Build GRCh38
rs864622648 Chr.2: 47377084-47377084 on Build GRCh38
rs772468246 Chr.2: 47376976-47376976 on Build GRCh38
rs1053865733 Chr.2: 47376948-47376948 on Build GRCh38
rs780740347 Chr.2: 47377112-47377112 on Build GRCh38
rs141632294 Chr.2: 47376979-47376980 on Build GRCh38
rs146685071 Chr.2: 47377015-47377015 on Build GRCh38
rs878854491 Chr.2: 47377045-47377045 on Build GRCh38
rs587780768 Chr.2: 47377018-47377018 on Build GRCh38
rs769475823 Chr.2: 47376989-47376989 on Build GRCh38
rs775172331 Chr.2: 47376991-47376991 on Build GRCh38
rs750348819 Chr.2: 47376967-47376967 on Build GRCh38
rs777222396 Chr.2: 47377054-47377054 on Build GRCh38
rs542716105 Chr.2: 47377006-47377006 on Build GRCh38
rs754496546 Chr.2: 47377036-47377036 on Build GRCh38
rs762488504 Chr.2: 47376996-47376996 on Build GRCh38
rs568235222 Chr.2: 47376957-47376957 on Build GRCh38
rs768280387 Chr.2: 47377073-47377073 on Build GRCh38
rs114020039 Chr.2: 47377153-47377153 on Build GRCh38
rs773087779 Chr.2: 47377116-47377116 on Build GRCh38
rs546921694 Chr.2: 47376955-47376955 on Build GRCh38
rs606231204 Chr.2: 47377021-47377022 on Build GRCh38
rs754990754 Chr.2: 47377023-47377023 on Build GRCh38
rs895260896 Chr.2: 47376975-47376975 on Build GRCh38
rs1042651324 Chr.2: 47377146-47377146 on Build GRCh38
rs199731885 Chr.2: 47377013-47377013 on Build GRCh38
rs780070922 Chr.2: 47376979-47376979 on Build GRCh38
rs606231281 Chr.2: 47377012-47377012 on Build GRCh38
rs772471530 Chr.2: 47377055-47377055 on Build GRCh38
rs912692525 Chr.2: 47377134-47377134 on Build GRCh38
rs78608315 Chr.2: 47377009-47377009 on Build GRCh38
rs4361175 Chr.2: 47377137-47377137 on Build GRCh38
rs987245397 Chr.2: 47377124-47377124 on Build GRCh38
rs780919846 Chr.2: 47377084-47377085 on Build GRCh38
rs920862989 Chr.2: 47377028-47377028 on Build GRCh38
rs754403432 Chr.2: 47376985-47376986 on Build GRCh38
rs115536548 Chr.2: 47377139-47377139 on Build GRCh38
rs939350013 Chr.2: 47377111-47377111 on Build GRCh38
rs557688130 Chr.2: 47377068-47377068 on Build GRCh38
rs751081548 Chr.2: 47376980-47376980 on Build GRCh38
rs763853364 Chr.2: 47377000-47377000 on Build GRCh38
rs761212041 Chr.2: 47376988-47376988 on Build GRCh38

View Full Product Details