Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 82356813-82357074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC59
Entrez Gene ID
29080
Gene Name
coiled-coil domain containing 59
Gene Aliases
BR22, HSPC128, TAP26
UniGene
Hs.582627
Gene Chromosome Location
Chr.12: 82352304-82358805 on Build GRCh38
Transcripts
NM_014167.4 NR_033192.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4647664 c.415T>G p.C139G Chr.12: 82357009-82357009 on Build GRCh38
5038513 c.359A>C p.H120P Chr.12: 82357065-82357065 on Build GRCh38
4641038 c.382T>C p.C128R Chr.12: 82357042-82357042 on Build GRCh38
4600037 c.427C>T p.Q143* Chr.12: 82356997-82356997 on Build GRCh38
372721 c.406G>A p.E136K Chr.12: 82357018-82357018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746930375 Chr.12: 82357040-82357040 on Build GRCh38
rs576655032 Chr.12: 82356863-82356863 on Build GRCh38
rs779915819 Chr.12: 82356935-82356935 on Build GRCh38
rs766749577 Chr.12: 82356970-82356970 on Build GRCh38
rs892362514 Chr.12: 82356965-82356965 on Build GRCh38
rs555201322 Chr.12: 82356862-82356862 on Build GRCh38
rs767520409 Chr.12: 82356991-82356991 on Build GRCh38
rs957658952 Chr.12: 82356844-82356844 on Build GRCh38
rs764109117 Chr.12: 82356949-82356949 on Build GRCh38
rs899656949 Chr.12: 82356903-82356903 on Build GRCh38
rs781269803 Chr.12: 82356914-82356914 on Build GRCh38
rs559579209 Chr.12: 82356964-82356964 on Build GRCh38
rs778715930 Chr.12: 82356918-82356918 on Build GRCh38
rs57682951 Chr.12: 82357003-82357003 on Build GRCh38
rs532659107 Chr.12: 82356948-82356948 on Build GRCh38
rs971891816 Chr.12: 82357000-82357000 on Build GRCh38
rs61737399 Chr.12: 82356992-82356992 on Build GRCh38
rs745703792 Chr.12: 82356920-82356920 on Build GRCh38
rs774728013 Chr.12: 82356984-82356984 on Build GRCh38
rs779139739 Chr.12: 82356957-82356966 on Build GRCh38
rs754891317 Chr.12: 82357042-82357042 on Build GRCh38
rs760033075 Chr.12: 82356987-82356987 on Build GRCh38
rs776148612 Chr.12: 82356958-82356958 on Build GRCh38
rs3751332 Chr.12: 82356845-82356845 on Build GRCh38
rs758312509 Chr.12: 82356930-82356930 on Build GRCh38
rs201456258 Chr.12: 82357038-82357038 on Build GRCh38
rs564330722 Chr.12: 82356962-82356962 on Build GRCh38
rs373386670 Chr.12: 82357010-82357010 on Build GRCh38
rs764352040 Chr.12: 82356997-82356997 on Build GRCh38
rs758258688 Chr.12: 82357031-82357031 on Build GRCh38
rs941308670 Chr.12: 82357022-82357022 on Build GRCh38
rs966118048 Chr.12: 82356890-82356890 on Build GRCh38
rs756159926 Chr.12: 82356993-82356993 on Build GRCh38
rs1000811419 Chr.12: 82356904-82356904 on Build GRCh38
rs1018198314 Chr.12: 82356866-82356866 on Build GRCh38
rs981548583 Chr.12: 82357012-82357012 on Build GRCh38
rs369283396 Chr.12: 82356967-82356967 on Build GRCh38
rs756284502 Chr.12: 82356915-82356915 on Build GRCh38

View Full Product Details