Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119300539-119300780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf3
Entrez Gene ID
401152
Gene Name
chromosome 4 open reading frame 3
Gene Aliases
-
UniGene
Hs.173705
Gene Chromosome Location
Chr.4: 119296419-119304445 on Build GRCh38
Transcripts
NM_001001701.3 NM_001170330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5568761 c.206C>G p.S69* Chr.4: 119300729-119300729 on Build GRCh38
3008061 c.306C>T p.A102A Chr.4: 119300629-119300629 on Build GRCh38
4649716 c.383G>C p.R128P Chr.4: 119300552-119300552 on Build GRCh38
7400366 c.307G>A p.G103R Chr.4: 119300628-119300628 on Build GRCh38
3825058 c.265T>G p.C89G Chr.4: 119300670-119300670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758474004 Chr.4: 119300572-119300572 on Build GRCh38
rs1039859411 Chr.4: 119300625-119300625 on Build GRCh38
rs779015240 Chr.4: 119300706-119300706 on Build GRCh38
rs765372959 Chr.4: 119300623-119300623 on Build GRCh38
rs745544034 Chr.4: 119300651-119300651 on Build GRCh38
rs761752242 Chr.4: 119300654-119300660 on Build GRCh38
rs973863509 Chr.4: 119300611-119300611 on Build GRCh38
rs763141591 Chr.4: 119300703-119300703 on Build GRCh38
rs771888106 Chr.4: 119300655-119300655 on Build GRCh38
rs566499624 Chr.4: 119300723-119300723 on Build GRCh38
rs183396708 Chr.4: 119300686-119300686 on Build GRCh38
rs373352889 Chr.4: 119300584-119300584 on Build GRCh38
rs764136208 Chr.4: 119300606-119300606 on Build GRCh38
rs746583722 Chr.4: 119300661-119300661 on Build GRCh38
rs754139603 Chr.4: 119300646-119300647 on Build GRCh38
rs753557076 Chr.4: 119300620-119300620 on Build GRCh38
rs779974911 Chr.4: 119300628-119300628 on Build GRCh38
rs776647539 Chr.4: 119300678-119300678 on Build GRCh38
rs779160155 Chr.4: 119300568-119300568 on Build GRCh38
rs765049962 Chr.4: 119300692-119300692 on Build GRCh38
rs369421902 Chr.4: 119300579-119300579 on Build GRCh38
rs751587404 Chr.4: 119300720-119300720 on Build GRCh38
rs868587311 Chr.4: 119300733-119300733 on Build GRCh38
rs553700594 Chr.4: 119300668-119300668 on Build GRCh38
rs756505658 Chr.4: 119300724-119300724 on Build GRCh38
rs1052436974 Chr.4: 119300719-119300719 on Build GRCh38
rs920006325 Chr.4: 119300748-119300748 on Build GRCh38
rs572065308 Chr.4: 119300560-119300560 on Build GRCh38
rs760608875 Chr.4: 119300641-119300642 on Build GRCh38
rs755040195 Chr.4: 119300593-119300593 on Build GRCh38
rs979669592 Chr.4: 119300709-119300709 on Build GRCh38
rs758422492 Chr.4: 119300626-119300626 on Build GRCh38
rs767000512 Chr.4: 119300576-119300576 on Build GRCh38
rs773033130 Chr.4: 119300696-119300696 on Build GRCh38
rs774836560 Chr.4: 119300658-119300658 on Build GRCh38
rs781752464 Chr.4: 119300636-119300636 on Build GRCh38
rs928604912 Chr.4: 119300639-119300639 on Build GRCh38
rs757444509 Chr.4: 119300566-119300566 on Build GRCh38
rs755090199 Chr.4: 119300721-119300721 on Build GRCh38
rs768416055 Chr.4: 119300672-119300672 on Build GRCh38
rs1023476604 Chr.4: 119300705-119300705 on Build GRCh38
rs1052810748 Chr.4: 119300652-119300652 on Build GRCh38
rs761519301 Chr.4: 119300581-119300581 on Build GRCh38
rs960458378 Chr.4: 119300699-119300699 on Build GRCh38
rs778658204 Chr.4: 119300645-119300645 on Build GRCh38
rs759050903 Chr.4: 119300596-119300596 on Build GRCh38
rs760510325 Chr.4: 119300600-119300600 on Build GRCh38
rs775451795 Chr.4: 119300599-119300599 on Build GRCh38
rs771741036 Chr.4: 119300598-119300598 on Build GRCh38
rs199807739 Chr.4: 119300578-119300578 on Build GRCh38
rs774161815 Chr.4: 119300589-119300589 on Build GRCh38
rs887349983 Chr.4: 119300674-119300674 on Build GRCh38
rs748455798 Chr.4: 119300637-119300637 on Build GRCh38
rs200591403 Chr.4: 119300629-119300629 on Build GRCh38
rs145398231 Chr.4: 119300722-119300722 on Build GRCh38
rs1025934760 Chr.4: 119300663-119300663 on Build GRCh38
rs748613373 Chr.4: 119300580-119300580 on Build GRCh38
rs942794162 Chr.4: 119300737-119300737 on Build GRCh38
rs569041917 Chr.4: 119300747-119300747 on Build GRCh38
rs755435031 Chr.4: 119300632-119300632 on Build GRCh38
rs750441719 Chr.4: 119300570-119300570 on Build GRCh38
rs770295559 Chr.4: 119300640-119300640 on Build GRCh38
rs527717324 Chr.4: 119300622-119300622 on Build GRCh38

View Full Product Details