Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64562268-64562507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A11
Entrez Gene ID
55867
Gene Name
solute carrier family 22 member 11
Gene Aliases
OAT4, hOAT4
UniGene
Hs.220844
Gene Chromosome Location
Chr.11: 64555601-64572875 on Build GRCh38
Transcripts
NM_001307985.1 NM_018484.3 XM_011545167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7204132 c.684G>A p.A228A Chr.11: 64562298-64562298 on Build GRCh38
2163871 c.685G>T p.V229F Chr.11: 64562299-64562299 on Build GRCh38
4726663 c.821G>T p.W274L Chr.11: 64562435-64562435 on Build GRCh38
4035156 c.679A>G p.R227G Chr.11: 64562293-64562293 on Build GRCh38
930133 c.709G>A p.A237T Chr.11: 64562323-64562323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371797963 Chr.11: 64562403-64562403 on Build GRCh38
rs751266040 Chr.11: 64562303-64562303 on Build GRCh38
rs1025497960 Chr.11: 64562328-64562328 on Build GRCh38
rs968352671 Chr.11: 64562367-64562367 on Build GRCh38
rs138025232 Chr.11: 64562353-64562353 on Build GRCh38
rs138399841 Chr.11: 64562342-64562342 on Build GRCh38
rs781501432 Chr.11: 64562310-64562310 on Build GRCh38
rs143517447 Chr.11: 64562431-64562431 on Build GRCh38
rs779595677 Chr.11: 64562372-64562372 on Build GRCh38
rs751377816 Chr.11: 64562364-64562364 on Build GRCh38
rs768654960 Chr.11: 64562477-64562477 on Build GRCh38
rs116034381 Chr.11: 64562309-64562309 on Build GRCh38
rs757063285 Chr.11: 64562371-64562371 on Build GRCh38
rs915102483 Chr.11: 64562402-64562402 on Build GRCh38
rs764766339 Chr.11: 64562352-64562352 on Build GRCh38
rs772492426 Chr.11: 64562440-64562440 on Build GRCh38
rs748672959 Chr.11: 64562435-64562435 on Build GRCh38
rs960776979 Chr.11: 64562378-64562378 on Build GRCh38
rs985289019 Chr.11: 64562360-64562360 on Build GRCh38
rs756379051 Chr.11: 64562381-64562381 on Build GRCh38
rs778080078 Chr.11: 64562444-64562444 on Build GRCh38
rs947933980 Chr.11: 64562297-64562297 on Build GRCh38
rs770582403 Chr.11: 64562346-64562346 on Build GRCh38
rs150160581 Chr.11: 64562332-64562332 on Build GRCh38
rs76833677 Chr.11: 64562298-64562298 on Build GRCh38
rs755510491 Chr.11: 64562430-64562430 on Build GRCh38
rs761840943 Chr.11: 64562483-64562483 on Build GRCh38
rs149251529 Chr.11: 64562349-64562349 on Build GRCh38
rs368497174 Chr.11: 64562343-64562343 on Build GRCh38
rs199681819 Chr.11: 64562482-64562482 on Build GRCh38
rs775099428 Chr.11: 64562469-64562469 on Build GRCh38
rs764713102 Chr.11: 64562293-64562293 on Build GRCh38
rs758987599 Chr.11: 64562292-64562292 on Build GRCh38
rs748391722 Chr.11: 64562345-64562345 on Build GRCh38
rs749284990 Chr.11: 64562406-64562406 on Build GRCh38
rs758237420 Chr.11: 64562295-64562295 on Build GRCh38
rs776121725 Chr.11: 64562285-64562285 on Build GRCh38
rs367925828 Chr.11: 64562331-64562331 on Build GRCh38
rs774446327 Chr.11: 64562413-64562420 on Build GRCh38
rs143422525 Chr.11: 64562351-64562351 on Build GRCh38
rs763899157 Chr.11: 64562355-64562355 on Build GRCh38
rs745570824 Chr.11: 64562453-64562453 on Build GRCh38
rs752696988 Chr.11: 64562294-64562294 on Build GRCh38
rs992205260 Chr.11: 64562417-64562417 on Build GRCh38
rs762304315 Chr.11: 64562470-64562470 on Build GRCh38

View Full Product Details