Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77130474-77130727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO7A
Entrez Gene ID
4647
Gene Name
myosin VIIA
Gene Aliases
DFNA11, DFNB2, MYOVIIA, MYU7A, NSRD2, USH1B
UniGene
Hs.370421
Gene Chromosome Location
Chr.11: 77128192-77215241 on Build GRCh38
Transcripts
NM_000260.3 NM_001127179.2 NM_001127180.1 XM_011545044.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7093448 c.10C>A p.L4I Chr.11: 77130644-77130644 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886048668 Chr.11: 77130615-77130615 on Build GRCh38
rs782582338 Chr.11: 77130700-77130700 on Build GRCh38
rs782511469 Chr.11: 77130673-77130673 on Build GRCh38
rs782628098 Chr.11: 77130563-77130563 on Build GRCh38
rs782787126 Chr.11: 77130637-77130637 on Build GRCh38
rs924061948 Chr.11: 77130529-77130529 on Build GRCh38
rs922150458 Chr.11: 77130577-77130577 on Build GRCh38
rs781844094 Chr.11: 77130671-77130671 on Build GRCh38
rs782357935 Chr.11: 77130624-77130624 on Build GRCh38
rs781989229 Chr.11: 77130625-77130625 on Build GRCh38
rs782202349 Chr.11: 77130699-77130700 on Build GRCh38
rs370329263 Chr.11: 77130587-77130587 on Build GRCh38
rs976111568 Chr.11: 77130576-77130576 on Build GRCh38
rs564622720 Chr.11: 77130654-77130654 on Build GRCh38
rs749574169 Chr.11: 77130618-77130618 on Build GRCh38
rs782044983 Chr.11: 77130667-77130667 on Build GRCh38
rs781942154 Chr.11: 77130608-77130608 on Build GRCh38
rs528175173 Chr.11: 77130516-77130516 on Build GRCh38
rs189611519 Chr.11: 77130691-77130691 on Build GRCh38
rs797044518 Chr.11: 77130635-77130635 on Build GRCh38
rs782126604 Chr.11: 77130626-77130626 on Build GRCh38
rs1036517538 Chr.11: 77130699-77130699 on Build GRCh38
rs544675846 Chr.11: 77130596-77130596 on Build GRCh38
rs782727289 Chr.11: 77130669-77130669 on Build GRCh38
rs782552620 Chr.11: 77130696-77130696 on Build GRCh38
rs782264432 Chr.11: 77130675-77130675 on Build GRCh38
rs782077926 Chr.11: 77130616-77130616 on Build GRCh38

View Full Product Details