Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35424248-35424501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPARD
Entrez Gene ID
5467
Gene Name
peroxisome proliferator activated receptor delta
Gene Aliases
FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB
UniGene
Hs.696032
Gene Chromosome Location
Chr.6: 35342558-35428191 on Build GRCh38
Transcripts
NM_001171818.1 NM_001171819.1 NM_001171820.1 NM_006238.4 NM_177435.2 XM_005249193.1 XM_006715120.1 XM_006715123.1 XM_011514707.1 XM_011514709.1 XM_011514710.1 XM_017010971.1 XM_017010972.1 XM_017010973.1 XM_017010974.1 XM_017010975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3994910 c.654A>C p.T218T Chr.6: 35424355-35424355 on Build GRCh38
4771905 c.637A>T p.I213F Chr.6: 35424338-35424338 on Build GRCh38
6529775 c.684G>C p.W228C Chr.6: 35424385-35424385 on Build GRCh38
3625998 c.709C>T p.P237S Chr.6: 35424410-35424410 on Build GRCh38
5357134 c.648C>T p.I216I Chr.6: 35424349-35424349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745661333 Chr.6: 35424279-35424279 on Build GRCh38
rs199521532 Chr.6: 35424469-35424469 on Build GRCh38
rs143029754 Chr.6: 35424376-35424376 on Build GRCh38
rs755176055 Chr.6: 35424283-35424283 on Build GRCh38
rs146127226 Chr.6: 35424396-35424396 on Build GRCh38
rs987573535 Chr.6: 35424424-35424424 on Build GRCh38
rs377039242 Chr.6: 35424312-35424312 on Build GRCh38
rs201479741 Chr.6: 35424349-35424349 on Build GRCh38
rs770202154 Chr.6: 35424430-35424430 on Build GRCh38
rs201419456 Chr.6: 35424443-35424443 on Build GRCh38
rs202166688 Chr.6: 35424382-35424382 on Build GRCh38
rs374511678 Chr.6: 35424433-35424433 on Build GRCh38
rs761749779 Chr.6: 35424344-35424344 on Build GRCh38
rs758550159 Chr.6: 35424403-35424403 on Build GRCh38
rs746220231 Chr.6: 35424427-35424427 on Build GRCh38
rs200496881 Chr.6: 35424434-35424434 on Build GRCh38
rs531223897 Chr.6: 35424317-35424317 on Build GRCh38
rs778414812 Chr.6: 35424295-35424295 on Build GRCh38
rs747075223 Chr.6: 35424406-35424406 on Build GRCh38
rs527545065 Chr.6: 35424272-35424272 on Build GRCh38
rs9658162 Chr.6: 35424313-35424313 on Build GRCh38
rs771421269 Chr.6: 35424410-35424410 on Build GRCh38
rs569276253 Chr.6: 35424269-35424269 on Build GRCh38
rs779312482 Chr.6: 35424288-35424288 on Build GRCh38
rs200917813 Chr.6: 35424289-35424289 on Build GRCh38
rs762434552 Chr.6: 35424316-35424316 on Build GRCh38
rs759841599 Chr.6: 35424310-35424310 on Build GRCh38
rs755905723 Chr.6: 35424352-35424352 on Build GRCh38
rs961795320 Chr.6: 35424462-35424462 on Build GRCh38
rs781582552 Chr.6: 35424412-35424412 on Build GRCh38
rs200779718 Chr.6: 35424473-35424473 on Build GRCh38
rs377438407 Chr.6: 35424440-35424440 on Build GRCh38
rs201754940 Chr.6: 35424318-35424318 on Build GRCh38
rs773800049 Chr.6: 35424460-35424460 on Build GRCh38
rs200192364 Chr.6: 35424343-35424343 on Build GRCh38
rs771349213 Chr.6: 35424301-35424301 on Build GRCh38
rs747620078 Chr.6: 35424296-35424296 on Build GRCh38
rs750380211 Chr.6: 35424474-35424474 on Build GRCh38
rs777935481 Chr.6: 35424404-35424404 on Build GRCh38
rs776996161 Chr.6: 35424307-35424307 on Build GRCh38

View Full Product Details