Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 152989926-152990151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTRF1L
Entrez Gene ID
54516
Gene Name
mitochondrial translational release factor 1 like
Gene Aliases
HMRF1L, MRF1L, mtRF1a
UniGene
Hs.225836
Gene Chromosome Location
Chr.6: 152987349-153007301 on Build GRCh38
Transcripts
NM_001114184.2 NM_001301047.2 NM_001301870.1 NM_001301871.1 NM_019041.6 NR_126056.1 XM_017010959.1 XM_017010960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4703668 c.1099G>A p.A367T Chr.6: 152989939-152989939 on Build GRCh38
3859783 c.1091A>G p.K364R Chr.6: 152989947-152989947 on Build GRCh38
1075181 c.1098C>T p.Y366Y Chr.6: 152989940-152989940 on Build GRCh38
740354 c.1089G>A p.L363L Chr.6: 152989949-152989949 on Build GRCh38
6846243 c.989C>T p.P330L Chr.6: 152990049-152990049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151055997 Chr.6: 152990076-152990076 on Build GRCh38
rs766785939 Chr.6: 152990056-152990059 on Build GRCh38
rs749206208 Chr.6: 152990091-152990091 on Build GRCh38
rs762100385 Chr.6: 152990029-152990029 on Build GRCh38
rs775321244 Chr.6: 152990088-152990088 on Build GRCh38
rs762190365 Chr.6: 152990101-152990101 on Build GRCh38
rs774943067 Chr.6: 152989956-152989957 on Build GRCh38
rs770754126 Chr.6: 152989954-152989954 on Build GRCh38
rs142813473 Chr.6: 152990043-152990043 on Build GRCh38
rs978639925 Chr.6: 152990124-152990124 on Build GRCh38
rs776268361 Chr.6: 152989958-152989958 on Build GRCh38
rs114354343 Chr.6: 152990103-152990103 on Build GRCh38
rs774555133 Chr.6: 152990098-152990098 on Build GRCh38
rs751869718 Chr.6: 152990013-152990013 on Build GRCh38
rs763146230 Chr.6: 152990003-152990003 on Build GRCh38
rs568513948 Chr.6: 152989983-152989983 on Build GRCh38
rs201452521 Chr.6: 152990064-152990064 on Build GRCh38
rs769616447 Chr.6: 152989984-152989984 on Build GRCh38
rs201682355 Chr.6: 152990046-152990046 on Build GRCh38
rs148517090 Chr.6: 152990007-152990007 on Build GRCh38
rs750746290 Chr.6: 152990032-152990032 on Build GRCh38
rs745661552 Chr.6: 152990083-152990083 on Build GRCh38
rs757937919 Chr.6: 152990059-152990059 on Build GRCh38
rs769580816 Chr.6: 152990086-152990086 on Build GRCh38
rs145697730 Chr.6: 152989993-152989993 on Build GRCh38
rs762415089 Chr.6: 152989989-152989993 on Build GRCh38
rs777235446 Chr.6: 152990061-152990061 on Build GRCh38
rs773429599 Chr.6: 152990121-152990121 on Build GRCh38
rs768903076 Chr.6: 152990095-152990095 on Build GRCh38
rs767980441 Chr.6: 152990030-152990030 on Build GRCh38
rs756437015 Chr.6: 152990042-152990042 on Build GRCh38

View Full Product Details