Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125084101-125084366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC37A2
Entrez Gene ID
219855
Gene Name
solute carrier family 37 member 2
Gene Aliases
pp11662
UniGene
Hs.352661
Gene Chromosome Location
Chr.11: 125063067-125090516 on Build GRCh38
Transcripts
NM_001145290.1 NM_198277.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1152270 c.1042G>A p.G348S Chr.11: 125084236-125084236 on Build GRCh38
686520 c.1060G>A p.V354I Chr.11: 125084254-125084254 on Build GRCh38
5646202 c.1107C>T p.L369L Chr.11: 125084301-125084301 on Build GRCh38
924504 c.1042G>A p.G348S Chr.11: 125084236-125084236 on Build GRCh38
5646201 c.1107C>T p.L369L Chr.11: 125084301-125084301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372521619 Chr.11: 125084324-125084324 on Build GRCh38
rs796316313 Chr.11: 125084291-125084291 on Build GRCh38
rs149511422 Chr.11: 125084272-125084272 on Build GRCh38
rs774075649 Chr.11: 125084213-125084213 on Build GRCh38
rs745343243 Chr.11: 125084207-125084207 on Build GRCh38
rs138434263 Chr.11: 125084274-125084274 on Build GRCh38
rs376182528 Chr.11: 125084337-125084337 on Build GRCh38
rs200568702 Chr.11: 125084254-125084254 on Build GRCh38
rs1005051641 Chr.11: 125084323-125084323 on Build GRCh38
rs779651234 Chr.11: 125084339-125084339 on Build GRCh38
rs527354827 Chr.11: 125084212-125084212 on Build GRCh38
rs532962515 Chr.11: 125084334-125084334 on Build GRCh38
rs73625362 Chr.11: 125084137-125084137 on Build GRCh38
rs764386708 Chr.11: 125084189-125084189 on Build GRCh38
rs923800697 Chr.11: 125084133-125084133 on Build GRCh38
rs747826796 Chr.11: 125084334-125084334 on Build GRCh38
rs776517414 Chr.11: 125084231-125084231 on Build GRCh38
rs1053666328 Chr.11: 125084153-125084153 on Build GRCh38
rs769596443 Chr.11: 125084252-125084252 on Build GRCh38
rs561672724 Chr.11: 125084180-125084180 on Build GRCh38
rs943180766 Chr.11: 125084242-125084242 on Build GRCh38
rs770700723 Chr.11: 125084201-125084201 on Build GRCh38
rs987159703 Chr.11: 125084132-125084132 on Build GRCh38
rs1044414586 Chr.11: 125084262-125084262 on Build GRCh38
rs199871161 Chr.11: 125084333-125084333 on Build GRCh38
rs775563553 Chr.11: 125084270-125084270 on Build GRCh38
rs374822921 Chr.11: 125084197-125084197 on Build GRCh38
rs756335995 Chr.11: 125084184-125084184 on Build GRCh38
rs746591573 Chr.11: 125084223-125084223 on Build GRCh38
rs751135963 Chr.11: 125084345-125084345 on Build GRCh38
rs368970945 Chr.11: 125084316-125084316 on Build GRCh38
rs776072731 Chr.11: 125084312-125084312 on Build GRCh38
rs1035763392 Chr.11: 125084307-125084307 on Build GRCh38
rs754167512 Chr.11: 125084281-125084281 on Build GRCh38
rs535796071 Chr.11: 125084236-125084236 on Build GRCh38
rs768433913 Chr.11: 125084224-125084224 on Build GRCh38
rs767570888 Chr.11: 125084260-125084260 on Build GRCh38
rs545427409 Chr.11: 125084289-125084289 on Build GRCh38
rs765646680 Chr.11: 125084293-125084293 on Build GRCh38
rs971740551 Chr.11: 125084126-125084126 on Build GRCh38
rs201412646 Chr.11: 125084347-125084347 on Build GRCh38
rs372222317 Chr.11: 125084196-125084196 on Build GRCh38
rs893725441 Chr.11: 125084171-125084171 on Build GRCh38
rs577943913 Chr.11: 125084164-125084164 on Build GRCh38
rs73024323 Chr.11: 125084346-125084346 on Build GRCh38

View Full Product Details