Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56716090-56716315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACA
Entrez Gene ID
4666
Gene Name
nascent polypeptide-associated complex alpha subunit
Gene Aliases
HSD48, NAC-alpha, NACA1, skNAC
UniGene
Hs.505735
Gene Chromosome Location
Chr.12: 56712427-56726275 on Build GRCh38
Transcripts
NM_001113201.2 NM_001113202.1 NM_001113203.2 NM_001320193.1 NM_001320194.1 NM_005594.5 XM_006719412.1 XM_006719413.2 XM_006719414.1 XM_006719418.1 XM_006719420.3 XM_011538389.1 XM_017019337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4147302 c.1924T>A p.S642T Chr.12: 56716147-56716147 on Build GRCh38
3359849 c.5217C>T p.L1739L Chr.12: 56716313-56716313 on Build GRCh38
5481077 c.1865-2A>C p.? Chr.12: 56716208-56716208 on Build GRCh38
5457544 c.5372_5373delCT p.S1791fs*22 Chr.12: 56716157-56716158 on Build GRCh38
5457543 c.1913_1914delCT p.S638fs*22 Chr.12: 56716157-56716158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371952741 Chr.12: 56716165-56716165 on Build GRCh38
rs745713255 Chr.12: 56716172-56716172 on Build GRCh38
rs530427863 Chr.12: 56716230-56716230 on Build GRCh38
rs755261178 Chr.12: 56716120-56716120 on Build GRCh38
rs773861647 Chr.12: 56716213-56716213 on Build GRCh38
rs888451523 Chr.12: 56716219-56716219 on Build GRCh38
rs983321183 Chr.12: 56716185-56716185 on Build GRCh38
rs368947398 Chr.12: 56716224-56716224 on Build GRCh38
rs986285206 Chr.12: 56716167-56716167 on Build GRCh38
rs541284931 Chr.12: 56716164-56716164 on Build GRCh38
rs145648145 Chr.12: 56716118-56716118 on Build GRCh38
rs187514929 Chr.12: 56716239-56716239 on Build GRCh38
rs766811694 Chr.12: 56716117-56716117 on Build GRCh38
rs763580591 Chr.12: 56716111-56716111 on Build GRCh38
rs781237360 Chr.12: 56716246-56716246 on Build GRCh38
rs372484247 Chr.12: 56716209-56716209 on Build GRCh38
rs754796663 Chr.12: 56716143-56716143 on Build GRCh38
rs914922212 Chr.12: 56716128-56716128 on Build GRCh38
rs749020893 Chr.12: 56716258-56716258 on Build GRCh38
rs769697215 Chr.12: 56716252-56716252 on Build GRCh38
rs919073619 Chr.12: 56716198-56716198 on Build GRCh38
rs2926747 Chr.12: 56716147-56716147 on Build GRCh38
rs768316820 Chr.12: 56716288-56716288 on Build GRCh38
rs934432450 Chr.12: 56716279-56716279 on Build GRCh38
rs780950107 Chr.12: 56716166-56716166 on Build GRCh38
rs779239441 Chr.12: 56716135-56716135 on Build GRCh38
rs766354403 Chr.12: 56716131-56716132 on Build GRCh38
rs766905792 Chr.12: 56716217-56716217 on Build GRCh38
rs775269882 Chr.12: 56716189-56716189 on Build GRCh38
rs747020698 Chr.12: 56716148-56716148 on Build GRCh38
rs199723539 Chr.12: 56716151-56716151 on Build GRCh38
rs752844855 Chr.12: 56716143-56716143 on Build GRCh38
rs111651575 Chr.12: 56716177-56716177 on Build GRCh38
rs754270901 Chr.12: 56716223-56716223 on Build GRCh38
rs753031480 Chr.12: 56716226-56716226 on Build GRCh38
rs183828032 Chr.12: 56716216-56716216 on Build GRCh38
rs778085879 Chr.12: 56716232-56716232 on Build GRCh38
rs924354237 Chr.12: 56716275-56716275 on Build GRCh38
rs745779934 Chr.12: 56716247-56716247 on Build GRCh38
rs751562955 Chr.12: 56716136-56716137 on Build GRCh38
rs757379727 Chr.12: 56716243-56716243 on Build GRCh38
rs570469875 Chr.12: 56716253-56716253 on Build GRCh38
rs200794039 Chr.12: 56716119-56716119 on Build GRCh38
rs149326268 Chr.12: 56716195-56716195 on Build GRCh38

View Full Product Details