Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247948798-247949043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2L13
Entrez Gene ID
284521
Gene Name
olfactory receptor family 2 subfamily L member 13
Gene Aliases
OR2L14
UniGene
Hs.372936
Gene Chromosome Location
Chr.1: 247937029-248100922 on Build GRCh38
Transcripts
NM_001304535.1 NM_175911.3 NM_001001963.1

Gene Symbol
OR2L8
Entrez Gene ID
391190
Gene Name
olfactory receptor family 2 subfamily L member 8 (gene/pseudogene)
Gene Aliases
-
UniGene
Hs.690212
Gene Chromosome Location
Chr.1: 247948858-247949796 on Build GRCh38
Transcripts
NM_001304535.1 NM_175911.3 NM_001001963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2232741 c.56C>T p.P19L Chr.1: 247948913-247948913 on Build GRCh38
5698999 c.156C>A p.T52T Chr.1: 247949013-247949013 on Build GRCh38
7381360 c.47T>A p.L16Q Chr.1: 247948904-247948904 on Build GRCh38
6288295 c.155C>T p.T52I Chr.1: 247949012-247949012 on Build GRCh38
2232740 c.8A>T p.N3I Chr.1: 247948865-247948865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551341888 Chr.1: 247949000-247949000 on Build GRCh38
rs147539830 Chr.1: 247948897-247948897 on Build GRCh38
rs771526321 Chr.1: 247948926-247948926 on Build GRCh38
rs754484374 Chr.1: 247948999-247948999 on Build GRCh38
rs774593675 Chr.1: 247948933-247948933 on Build GRCh38
rs765055101 Chr.1: 247948951-247948951 on Build GRCh38
rs751077152 Chr.1: 247948998-247948998 on Build GRCh38
rs756488004 Chr.1: 247948912-247948912 on Build GRCh38
rs370313583 Chr.1: 247948878-247948878 on Build GRCh38
rs745883430 Chr.1: 247948935-247948935 on Build GRCh38
rs753587766 Chr.1: 247948855-247948855 on Build GRCh38
rs371286821 Chr.1: 247949010-247949010 on Build GRCh38
rs527380894 Chr.1: 247949013-247949013 on Build GRCh38
rs765150531 Chr.1: 247948868-247948868 on Build GRCh38
rs749772956 Chr.1: 247948925-247948925 on Build GRCh38
rs756476412 Chr.1: 247949008-247949008 on Build GRCh38
rs538787027 Chr.1: 247948891-247948891 on Build GRCh38
rs187754853 Chr.1: 247948832-247948832 on Build GRCh38
rs762902268 Chr.1: 247948980-247948980 on Build GRCh38
rs766249273 Chr.1: 247948985-247948985 on Build GRCh38
rs199564310 Chr.1: 247948835-247948835 on Build GRCh38
rs772426717 Chr.1: 247948941-247948941 on Build GRCh38
rs566489004 Chr.1: 247949002-247949002 on Build GRCh38
rs767797637 Chr.1: 247948853-247948853 on Build GRCh38
rs750357242 Chr.1: 247948869-247948869 on Build GRCh38
rs977362725 Chr.1: 247949009-247949009 on Build GRCh38
rs965813620 Chr.1: 247948972-247948972 on Build GRCh38
rs376181982 Chr.1: 247948967-247948967 on Build GRCh38
rs757949835 Chr.1: 247948871-247948871 on Build GRCh38
rs765841397 Chr.1: 247948875-247948875 on Build GRCh38
rs754642117 Chr.1: 247948895-247948895 on Build GRCh38
rs748532567 Chr.1: 247948908-247948908 on Build GRCh38
rs1027546990 Chr.1: 247948861-247948861 on Build GRCh38
rs921245905 Chr.1: 247948922-247948922 on Build GRCh38
rs774757116 Chr.1: 247948836-247948836 on Build GRCh38
rs761566482 Chr.1: 247948945-247948945 on Build GRCh38
rs761602515 Chr.1: 247948866-247948866 on Build GRCh38
rs747463100 Chr.1: 247948876-247948877 on Build GRCh38
rs372980688 Chr.1: 247948914-247948914 on Build GRCh38

View Full Product Details