Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7318482-7318711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEURL4
Entrez Gene ID
84461
Gene Name
neuralized E3 ubiquitin protein ligase 4
Gene Aliases
-
UniGene
Hs.654794
Gene Chromosome Location
Chr.17: 7315632-7329319 on Build GRCh38
Transcripts
NM_001005408.1 NM_032442.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1716192 c.3716C>T p.T1239M Chr.17: 7318643-7318643 on Build GRCh38
3521686 c.3759T>C p.S1253S Chr.17: 7318600-7318600 on Build GRCh38
5016537 c.3811_3830del20 p.D1271fs*10 Chr.17: 7318529-7318548 on Build GRCh38
4688313 c.3799delG p.V1267fs*1 Chr.17: 7318560-7318560 on Build GRCh38
3937547 c.3817C>A p.P1273T Chr.17: 7318536-7318536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765935159 Chr.17: 7318583-7318583 on Build GRCh38
rs753737221 Chr.17: 7318614-7318614 on Build GRCh38
rs777751368 Chr.17: 7318513-7318513 on Build GRCh38
rs371381411 Chr.17: 7318643-7318643 on Build GRCh38
rs755927839 Chr.17: 7318511-7318511 on Build GRCh38
rs369653182 Chr.17: 7318529-7318529 on Build GRCh38
rs763456916 Chr.17: 7318659-7318659 on Build GRCh38
rs753609480 Chr.17: 7318691-7318691 on Build GRCh38
rs747943314 Chr.17: 7318505-7318505 on Build GRCh38
rs754574034 Chr.17: 7318597-7318597 on Build GRCh38
rs1043881207 Chr.17: 7318678-7318678 on Build GRCh38
rs768848362 Chr.17: 7318528-7318528 on Build GRCh38
rs374635885 Chr.17: 7318653-7318653 on Build GRCh38
rs373069299 Chr.17: 7318633-7318633 on Build GRCh38
rs200948360 Chr.17: 7318669-7318669 on Build GRCh38
rs760278963 Chr.17: 7318689-7318689 on Build GRCh38
rs374468101 Chr.17: 7318541-7318541 on Build GRCh38
rs973273139 Chr.17: 7318548-7318548 on Build GRCh38
rs752348342 Chr.17: 7318604-7318604 on Build GRCh38
rs201919127 Chr.17: 7318624-7318624 on Build GRCh38
rs765580978 Chr.17: 7318560-7318560 on Build GRCh38
rs776763243 Chr.17: 7318531-7318531 on Build GRCh38
rs775064128 Chr.17: 7318668-7318668 on Build GRCh38
rs200344128 Chr.17: 7318690-7318690 on Build GRCh38
rs762235879 Chr.17: 7318552-7318552 on Build GRCh38
rs777436315 Chr.17: 7318612-7318612 on Build GRCh38
rs370681631 Chr.17: 7318666-7318666 on Build GRCh38
rs747064051 Chr.17: 7318630-7318630 on Build GRCh38
rs773730980 Chr.17: 7318656-7318656 on Build GRCh38
rs377003323 Chr.17: 7318521-7318521 on Build GRCh38
rs201910864 Chr.17: 7318526-7318526 on Build GRCh38
rs750468168 Chr.17: 7318660-7318660 on Build GRCh38
rs778871451 Chr.17: 7318625-7318625 on Build GRCh38
rs1052575933 Chr.17: 7318661-7318661 on Build GRCh38
rs755878935 Chr.17: 7318605-7318605 on Build GRCh38
rs763583170 Chr.17: 7318572-7318572 on Build GRCh38
rs374233802 Chr.17: 7318591-7318591 on Build GRCh38
rs773699370 Chr.17: 7318561-7318561 on Build GRCh38
rs767194830 Chr.17: 7318599-7318599 on Build GRCh38
rs552299123 Chr.17: 7318682-7318682 on Build GRCh38
rs767144385 Chr.17: 7318576-7318576 on Build GRCh38
rs117984863 Chr.17: 7318670-7318670 on Build GRCh38
rs775578113 Chr.17: 7318525-7318525 on Build GRCh38
rs376177328 Chr.17: 7318642-7318642 on Build GRCh38
rs1026566717 Chr.17: 7318567-7318567 on Build GRCh38

View Full Product Details