Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75635749-75635994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KARS
Entrez Gene ID
3735
Gene Name
lysyl-tRNA synthetase
Gene Aliases
CMTRIB, DFNB89, KARS1, KARS2, KRS
UniGene
Hs.3100
Gene Chromosome Location
Chr.16: 75627724-75647687 on Build GRCh38
Transcripts
NM_001130089.1 NM_005548.2 XM_017023217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3512283 c.682C>T p.P228S Chr.16: 75635983-75635983 on Build GRCh38
4062872 c.766C>T p.R256C Chr.16: 75635793-75635793 on Build GRCh38
3402482 c.684G>A p.P228P Chr.16: 75635981-75635981 on Build GRCh38
2155406 c.691T>C p.Y231H Chr.16: 75635784-75635784 on Build GRCh38
973763 c.636G>C p.M212I Chr.16: 75635945-75635945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150091114 Chr.16: 75635797-75635797 on Build GRCh38
rs768730599 Chr.16: 75635827-75635827 on Build GRCh38
rs200848719 Chr.16: 75635968-75635968 on Build GRCh38
rs574836321 Chr.16: 75635793-75635793 on Build GRCh38
rs776073313 Chr.16: 75635878-75635880 on Build GRCh38
rs761711998 Chr.16: 75635783-75635783 on Build GRCh38
rs968882904 Chr.16: 75635892-75635892 on Build GRCh38
rs762348610 Chr.16: 75635958-75635958 on Build GRCh38
rs759875793 Chr.16: 75635799-75635799 on Build GRCh38
rs762878161 Chr.16: 75635877-75635877 on Build GRCh38
rs149424412 Chr.16: 75635963-75635963 on Build GRCh38
rs774089333 Chr.16: 75635781-75635781 on Build GRCh38
rs751637676 Chr.16: 75635947-75635947 on Build GRCh38
rs763110390 Chr.16: 75635908-75635908 on Build GRCh38
rs553641161 Chr.16: 75635860-75635860 on Build GRCh38
rs767476599 Chr.16: 75635792-75635792 on Build GRCh38
rs141539149 Chr.16: 75635969-75635969 on Build GRCh38
rs781604856 Chr.16: 75635854-75635854 on Build GRCh38
rs369950194 Chr.16: 75635895-75635895 on Build GRCh38
rs755841211 Chr.16: 75635875-75635875 on Build GRCh38
rs757751625 Chr.16: 75635845-75635845 on Build GRCh38
rs201237548 Chr.16: 75635800-75635800 on Build GRCh38
rs773035996 Chr.16: 75635849-75635849 on Build GRCh38
rs772621409 Chr.16: 75635874-75635887 on Build GRCh38
rs181461914 Chr.16: 75635865-75635865 on Build GRCh38
rs964697338 Chr.16: 75635841-75635841 on Build GRCh38
rs201206783 Chr.16: 75635890-75635890 on Build GRCh38
rs80068681 Chr.16: 75635836-75635836 on Build GRCh38
rs138927699 Chr.16: 75635945-75635945 on Build GRCh38
rs750755615 Chr.16: 75635869-75635869 on Build GRCh38
rs572731802 Chr.16: 75635879-75635879 on Build GRCh38
rs748163106 Chr.16: 75635889-75635889 on Build GRCh38
rs764142658 Chr.16: 75635923-75635923 on Build GRCh38
rs200576782 Chr.16: 75635810-75635810 on Build GRCh38
rs767991207 Chr.16: 75635881-75635881 on Build GRCh38
rs761020489 Chr.16: 75635960-75635964 on Build GRCh38
rs760336235 Chr.16: 75635896-75635896 on Build GRCh38
rs762905187 Chr.16: 75635817-75635817 on Build GRCh38
rs72789443 Chr.16: 75635906-75635906 on Build GRCh38
rs375607294 Chr.16: 75635815-75635815 on Build GRCh38
rs778552223 Chr.16: 75635882-75635882 on Build GRCh38
rs201821932 Chr.16: 75635922-75635922 on Build GRCh38
rs372430772 Chr.16: 75635905-75635905 on Build GRCh38

View Full Product Details