Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1697517-1697768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP5-6
Entrez Gene ID
440023
Gene Name
keratin associated protein 5-6
Gene Aliases
KRTAP5.6
UniGene
Hs.534863
Gene Chromosome Location
Chr.11: 1697195-1697755 on Build GRCh38
Transcripts
NM_001012416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1352908 c.385A>C p.I129L Chr.11: 1697630-1697630 on Build GRCh38
925449 c.311G>A p.C104Y Chr.11: 1697556-1697556 on Build GRCh38
4802895 c.345C>T p.S115S Chr.11: 1697590-1697590 on Build GRCh38
3676138 c.289T>C p.S97P Chr.11: 1697534-1697534 on Build GRCh38
7257452 c.349G>T p.A117S Chr.11: 1697594-1697594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751437617 Chr.11: 1697678-1697678 on Build GRCh38
rs759610168 Chr.11: 1697562-1697562 on Build GRCh38
rs189317877 Chr.11: 1697626-1697626 on Build GRCh38
rs773631793 Chr.11: 1697670-1697670 on Build GRCh38
rs533880875 Chr.11: 1697722-1697722 on Build GRCh38
rs181311214 Chr.11: 1697663-1697663 on Build GRCh38
rs765408057 Chr.11: 1697567-1697567 on Build GRCh38
rs148475449 Chr.11: 1697623-1697623 on Build GRCh38
rs779667447 Chr.11: 1697614-1697614 on Build GRCh38
rs185268567 Chr.11: 1697710-1697710 on Build GRCh38
rs761078059 Chr.11: 1697542-1697542 on Build GRCh38
rs527835026 Chr.11: 1697551-1697551 on Build GRCh38
rs758283177 Chr.11: 1697578-1697578 on Build GRCh38
rs539426314 Chr.11: 1697695-1697695 on Build GRCh38
rs775237593 Chr.11: 1697536-1697536 on Build GRCh38
rs373157622 Chr.11: 1697675-1697675 on Build GRCh38
rs756996891 Chr.11: 1697596-1697596 on Build GRCh38
rs893652864 Chr.11: 1697653-1697653 on Build GRCh38
rs925763671 Chr.11: 1697711-1697711 on Build GRCh38
rs747612088 Chr.11: 1697656-1697656 on Build GRCh38
rs752805481 Chr.11: 1697571-1697571 on Build GRCh38
rs372985780 Chr.11: 1697541-1697541 on Build GRCh38
rs572009313 Chr.11: 1697673-1697673 on Build GRCh38
rs750678767 Chr.11: 1697623-1697624 on Build GRCh38
rs201960565 Chr.11: 1697661-1697661 on Build GRCh38
rs188524664 Chr.11: 1697737-1697737 on Build GRCh38
rs557719152 Chr.11: 1697700-1697700 on Build GRCh38
rs73404785 Chr.11: 1697603-1697603 on Build GRCh38
rs763298292 Chr.11: 1697554-1697554 on Build GRCh38
rs767769152 Chr.11: 1697545-1697557 on Build GRCh38
rs765467621 Chr.11: 1697667-1697667 on Build GRCh38
rs757266125 Chr.11: 1697684-1697684 on Build GRCh38
rs762302872 Chr.11: 1697540-1697540 on Build GRCh38
rs773854084 Chr.11: 1697637-1697637 on Build GRCh38
rs568835250 Chr.11: 1697669-1697669 on Build GRCh38
rs753875064 Chr.11: 1697556-1697556 on Build GRCh38
rs767183096 Chr.11: 1697686-1697686 on Build GRCh38
rs777095981 Chr.11: 1697662-1697662 on Build GRCh38
rs751287614 Chr.11: 1697593-1697593 on Build GRCh38
rs777701061 Chr.11: 1697586-1697586 on Build GRCh38
rs781097573 Chr.11: 1697598-1697598 on Build GRCh38
rs745483857 Chr.11: 1697602-1697602 on Build GRCh38

View Full Product Details