Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60773331-60773556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A15
Entrez Gene ID
219995
Gene Name
membrane spanning 4-domains A15
Gene Aliases
-
UniGene
Hs.207465
Gene Chromosome Location
Chr.11: 60756867-60776733 on Build GRCh38
Transcripts
NM_001098835.1 NM_001278242.1 NM_152717.2 NR_103481.1 XM_006718459.3 XM_011544812.2 XM_011544814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4034445 c.160G>A p.V54I Chr.11: 60773425-60773425 on Build GRCh38
4034444 c.439G>A p.V147I Chr.11: 60773425-60773425 on Build GRCh38
7226159 c.130A>T p.R44W Chr.11: 60773395-60773395 on Build GRCh38
3931363 c.150C>T p.N50N Chr.11: 60773415-60773415 on Build GRCh38
4401891 c.199G>A p.D67N Chr.11: 60773464-60773464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570606661 Chr.11: 60773432-60773432 on Build GRCh38
rs753312167 Chr.11: 60773423-60773423 on Build GRCh38
rs550872769 Chr.11: 60773431-60773431 on Build GRCh38
rs757388542 Chr.11: 60773362-60773362 on Build GRCh38
rs202147867 Chr.11: 60773366-60773366 on Build GRCh38
rs148137319 Chr.11: 60773483-60773483 on Build GRCh38
rs140880873 Chr.11: 60773400-60773400 on Build GRCh38
rs762028255 Chr.11: 60773505-60773505 on Build GRCh38
rs764469716 Chr.11: 60773498-60773498 on Build GRCh38
rs374382863 Chr.11: 60773506-60773506 on Build GRCh38
rs761572079 Chr.11: 60773398-60773398 on Build GRCh38
rs143783115 Chr.11: 60773434-60773434 on Build GRCh38
rs895606945 Chr.11: 60773393-60773393 on Build GRCh38
rs200595371 Chr.11: 60773502-60773502 on Build GRCh38
rs201539546 Chr.11: 60773435-60773435 on Build GRCh38
rs979685168 Chr.11: 60773444-60773444 on Build GRCh38
rs539829291 Chr.11: 60773433-60773433 on Build GRCh38
rs746938161 Chr.11: 60773374-60773374 on Build GRCh38
rs754969932 Chr.11: 60773526-60773526 on Build GRCh38
rs370909365 Chr.11: 60773386-60773386 on Build GRCh38
rs767242831 Chr.11: 60773363-60773376 on Build GRCh38
rs768507781 Chr.11: 60773382-60773382 on Build GRCh38
rs780921730 Chr.11: 60773532-60773532 on Build GRCh38
rs750714599 Chr.11: 60773507-60773507 on Build GRCh38
rs201439879 Chr.11: 60773411-60773411 on Build GRCh38
rs775349220 Chr.11: 60773370-60773370 on Build GRCh38
rs201653204 Chr.11: 60773484-60773484 on Build GRCh38
rs756683966 Chr.11: 60773424-60773424 on Build GRCh38
rs768149787 Chr.11: 60773422-60773422 on Build GRCh38
rs772003100 Chr.11: 60773369-60773369 on Build GRCh38
rs566525311 Chr.11: 60773470-60773470 on Build GRCh38
rs763108976 Chr.11: 60773521-60773521 on Build GRCh38
rs764559217 Chr.11: 60773425-60773425 on Build GRCh38
rs754103339 Chr.11: 60773428-60773428 on Build GRCh38
rs199752508 Chr.11: 60773488-60773488 on Build GRCh38
rs747853565 Chr.11: 60773473-60773473 on Build GRCh38
rs750081928 Chr.11: 60773521-60773521 on Build GRCh38
rs761313254 Chr.11: 60773489-60773489 on Build GRCh38
rs530687753 Chr.11: 60773415-60773415 on Build GRCh38
rs768459356 Chr.11: 60773462-60773462 on Build GRCh38
rs1030567644 Chr.11: 60773533-60773533 on Build GRCh38
rs757530633 Chr.11: 60773429-60773429 on Build GRCh38
rs557792793 Chr.11: 60773364-60773364 on Build GRCh38
rs780161146 Chr.11: 60773518-60773518 on Build GRCh38
rs772939478 Chr.11: 60773482-60773482 on Build GRCh38
rs964893824 Chr.11: 60773485-60773485 on Build GRCh38
rs758591099 Chr.11: 60773515-60773515 on Build GRCh38

View Full Product Details