Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105176521-105176774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT14
Entrez Gene ID
256281
Gene Name
nudix hydrolase 14
Gene Aliases
UGPP, UGPPase
UniGene
Hs.526432
Gene Chromosome Location
Chr.14: 105172937-105181323 on Build GRCh38
Transcripts
NM_001318380.1 NM_177533.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
697779 c.324G>T p.V108V Chr.14: 105176638-105176638 on Build GRCh38
459211 c.283G>A p.G95S Chr.14: 105176679-105176679 on Build GRCh38
7139834 c.230C>G p.S77C Chr.14: 105176732-105176732 on Build GRCh38
4613361 c.295delG p.V99fs*1 Chr.14: 105176667-105176667 on Build GRCh38
120976 c.259C>G p.R87G Chr.14: 105176703-105176703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117624322 Chr.14: 105176680-105176680 on Build GRCh38
rs760422307 Chr.14: 105176682-105176682 on Build GRCh38
rs768239721 Chr.14: 105176603-105176603 on Build GRCh38
rs780400619 Chr.14: 105176716-105176716 on Build GRCh38
rs754096722 Chr.14: 105176544-105176544 on Build GRCh38
rs748893001 Chr.14: 105176647-105176647 on Build GRCh38
rs587661158 Chr.14: 105176553-105176553 on Build GRCh38
rs748155368 Chr.14: 105176672-105176672 on Build GRCh38
rs778776773 Chr.14: 105176619-105176619 on Build GRCh38
rs375404228 Chr.14: 105176564-105176564 on Build GRCh38
rs1019049412 Chr.14: 105176700-105176700 on Build GRCh38
rs755152684 Chr.14: 105176549-105176549 on Build GRCh38
rs775112094 Chr.14: 105176584-105176584 on Build GRCh38
rs754665869 Chr.14: 105176721-105176721 on Build GRCh38
rs770714217 Chr.14: 105176698-105176698 on Build GRCh38
rs766286499 Chr.14: 105176649-105176650 on Build GRCh38
rs374599884 Chr.14: 105176739-105176739 on Build GRCh38
rs745598362 Chr.14: 105176580-105176580 on Build GRCh38
rs369128988 Chr.14: 105176582-105176582 on Build GRCh38
rs754734744 Chr.14: 105176709-105176710 on Build GRCh38
rs768842524 Chr.14: 105176754-105176754 on Build GRCh38
rs200578196 Chr.14: 105176671-105176671 on Build GRCh38
rs763216252 Chr.14: 105176752-105176752 on Build GRCh38
rs1020474342 Chr.14: 105176545-105176545 on Build GRCh38
rs587693886 Chr.14: 105176589-105176589 on Build GRCh38
rs761775506 Chr.14: 105176609-105176609 on Build GRCh38
rs751623123 Chr.14: 105176705-105176705 on Build GRCh38
rs150187377 Chr.14: 105176641-105176641 on Build GRCh38
rs776059102 Chr.14: 105176690-105176690 on Build GRCh38
rs746282769 Chr.14: 105176621-105176621 on Build GRCh38
rs148480421 Chr.14: 105176744-105176744 on Build GRCh38
rs371307451 Chr.14: 105176738-105176738 on Build GRCh38
rs753612516 Chr.14: 105176720-105176720 on Build GRCh38
rs144645678 Chr.14: 105176709-105176709 on Build GRCh38
rs1033656211 Chr.14: 105176578-105176578 on Build GRCh38
rs752063309 Chr.14: 105176640-105176640 on Build GRCh38
rs747463026 Chr.14: 105176561-105176561 on Build GRCh38
rs770632103 Chr.14: 105176745-105176745 on Build GRCh38
rs377703605 Chr.14: 105176719-105176719 on Build GRCh38
rs763091731 Chr.14: 105176702-105176702 on Build GRCh38
rs757778762 Chr.14: 105176707-105176707 on Build GRCh38
rs768286527 Chr.14: 105176668-105176668 on Build GRCh38
rs753903685 Chr.14: 105176578-105176578 on Build GRCh38
rs751512819 Chr.14: 105176680-105176680 on Build GRCh38
rs776403747 Chr.14: 105176748-105176748 on Build GRCh38
rs765418409 Chr.14: 105176550-105176550 on Build GRCh38
rs781341775 Chr.14: 105176571-105176571 on Build GRCh38
rs201161191 Chr.14: 105176576-105176576 on Build GRCh38
rs372965453 Chr.14: 105176610-105176610 on Build GRCh38
rs747633718 Chr.14: 105176734-105176734 on Build GRCh38
rs772115169 Chr.14: 105176678-105176678 on Build GRCh38
rs138979378 Chr.14: 105176646-105176646 on Build GRCh38
rs373290193 Chr.14: 105176552-105176552 on Build GRCh38
rs764223059 Chr.14: 105176703-105176703 on Build GRCh38
rs764099627 Chr.14: 105176637-105176637 on Build GRCh38
rs748716510 Chr.14: 105176601-105176601 on Build GRCh38
rs141847132 Chr.14: 105176679-105176679 on Build GRCh38
rs372614140 Chr.14: 105176627-105176627 on Build GRCh38
rs759854798 Chr.14: 105176620-105176620 on Build GRCh38
rs957735973 Chr.14: 105176593-105176593 on Build GRCh38
rs587616275 Chr.14: 105176649-105176649 on Build GRCh38
rs778286917 Chr.14: 105176556-105176556 on Build GRCh38
rs750901508 Chr.14: 105176712-105176712 on Build GRCh38
rs774444519 Chr.14: 105176607-105176607 on Build GRCh38
rs756456051 Chr.14: 105176713-105176713 on Build GRCh38
rs762354981 Chr.14: 105176706-105176706 on Build GRCh38
rs778617226 Chr.14: 105176730-105176730 on Build GRCh38
rs750730000 Chr.14: 105176644-105176644 on Build GRCh38

View Full Product Details