Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 80718720-80718969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYF5
Entrez Gene ID
4617
Gene Name
myogenic factor 5
Gene Aliases
bHLHc2
UniGene
Hs.178023
Gene Chromosome Location
Chr.12: 80716929-80719668 on Build GRCh38
Transcripts
NM_005593.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1993405 c.661G>T p.G221W Chr.12: 80718944-80718944 on Build GRCh38
1993406 c.662G>A p.G221E Chr.12: 80718945-80718945 on Build GRCh38
6138442 c.670C>A p.L224I Chr.12: 80718953-80718953 on Build GRCh38
6138443 c.654G>T p.E218D Chr.12: 80718937-80718937 on Build GRCh38
216290 c.637C>A p.R213R Chr.12: 80718920-80718920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143735700 Chr.12: 80718772-80718772 on Build GRCh38
rs546065836 Chr.12: 80718768-80718768 on Build GRCh38
rs1008856362 Chr.12: 80718809-80718809 on Build GRCh38
rs919999610 Chr.12: 80718771-80718771 on Build GRCh38
rs757528124 Chr.12: 80718867-80718867 on Build GRCh38
rs774372311 Chr.12: 80718921-80718921 on Build GRCh38
rs769714683 Chr.12: 80718820-80718820 on Build GRCh38
rs776991226 Chr.12: 80718911-80718911 on Build GRCh38
rs879130551 Chr.12: 80718822-80718822 on Build GRCh38
rs760144671 Chr.12: 80718807-80718807 on Build GRCh38
rs780215515 Chr.12: 80718892-80718892 on Build GRCh38
rs929784255 Chr.12: 80718783-80718783 on Build GRCh38
rs752147258 Chr.12: 80718949-80718949 on Build GRCh38
rs146104243 Chr.12: 80718907-80718907 on Build GRCh38
rs879201710 Chr.12: 80718885-80718885 on Build GRCh38
rs1045548045 Chr.12: 80718791-80718791 on Build GRCh38
rs763418336 Chr.12: 80718859-80718860 on Build GRCh38
rs772224652 Chr.12: 80718888-80718888 on Build GRCh38
rs780419800 Chr.12: 80718845-80718845 on Build GRCh38
rs955559054 Chr.12: 80718941-80718941 on Build GRCh38
rs759355407 Chr.12: 80718936-80718936 on Build GRCh38
rs766811135 Chr.12: 80718950-80718951 on Build GRCh38
rs773082739 Chr.12: 80718829-80718829 on Build GRCh38
rs372819612 Chr.12: 80718859-80718859 on Build GRCh38
rs759135661 Chr.12: 80718855-80718855 on Build GRCh38
rs779520098 Chr.12: 80718871-80718871 on Build GRCh38
rs180777844 Chr.12: 80718762-80718762 on Build GRCh38
rs563652605 Chr.12: 80718751-80718751 on Build GRCh38
rs773802546 Chr.12: 80718829-80718830 on Build GRCh38
rs751421525 Chr.12: 80718848-80718848 on Build GRCh38
rs762847236 Chr.12: 80718915-80718915 on Build GRCh38
rs755699591 Chr.12: 80718861-80718861 on Build GRCh38
rs1001234291 Chr.12: 80718802-80718802 on Build GRCh38
rs778127383 Chr.12: 80718863-80718863 on Build GRCh38
rs766892886 Chr.12: 80718856-80718856 on Build GRCh38
rs906920078 Chr.12: 80718792-80718792 on Build GRCh38
rs547514043 Chr.12: 80718909-80718909 on Build GRCh38
rs762830414 Chr.12: 80718842-80718842 on Build GRCh38
rs754265550 Chr.12: 80718865-80718865 on Build GRCh38
rs752382196 Chr.12: 80718857-80718857 on Build GRCh38
rs746308787 Chr.12: 80718883-80718883 on Build GRCh38
rs767020497 Chr.12: 80718944-80718944 on Build GRCh38
rs372785185 Chr.12: 80718920-80718920 on Build GRCh38
rs565120427 Chr.12: 80718813-80718813 on Build GRCh38
rs1022945666 Chr.12: 80718880-80718880 on Build GRCh38

View Full Product Details