Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112071133-112071360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIH1D2
Entrez Gene ID
120379
Gene Name
PIH1 domain containing 2
Gene Aliases
-
UniGene
Hs.420662
Gene Chromosome Location
Chr.11: 112052161-112074288 on Build GRCh38
Transcripts
NM_001082619.1 NM_138789.3 XM_011542593.2 XM_017017201.1 XM_017017202.1 XM_017017203.1 XM_017017204.1 XM_017017205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1726076 c.311C>A p.T104K Chr.11: 112071274-112071274 on Build GRCh38
5083356 c.377_378insA p.N126fs*12 Chr.11: 112071207-112071208 on Build GRCh38
5837801 c.377_378insA p.N126fs*12 Chr.11: 112071207-112071208 on Build GRCh38
5837800 c.393G>A p.M131I Chr.11: 112071192-112071192 on Build GRCh38
5012434 c.361G>C p.D121H Chr.11: 112071224-112071224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781989264 Chr.11: 112071284-112071284 on Build GRCh38
rs966633461 Chr.11: 112071262-112071262 on Build GRCh38
rs990246407 Chr.11: 112071297-112071297 on Build GRCh38
rs782186801 Chr.11: 112071194-112071194 on Build GRCh38
rs1033882124 Chr.11: 112071193-112071193 on Build GRCh38
rs782082491 Chr.11: 112071250-112071250 on Build GRCh38
rs782303101 Chr.11: 112071224-112071224 on Build GRCh38
rs79969687 Chr.11: 112071280-112071280 on Build GRCh38
rs782145459 Chr.11: 112071321-112071321 on Build GRCh38
rs782036048 Chr.11: 112071240-112071240 on Build GRCh38
rs1425917 Chr.11: 112071235-112071235 on Build GRCh38
rs977784722 Chr.11: 112071264-112071264 on Build GRCh38
rs782492751 Chr.11: 112071165-112071165 on Build GRCh38
rs1024887283 Chr.11: 112071247-112071247 on Build GRCh38
rs587726153 Chr.11: 112071178-112071178 on Build GRCh38
rs924641159 Chr.11: 112071266-112071266 on Build GRCh38
rs151247493 Chr.11: 112071181-112071181 on Build GRCh38
rs959191888 Chr.11: 112071233-112071233 on Build GRCh38
rs782257962 Chr.11: 112071207-112071208 on Build GRCh38
rs887486506 Chr.11: 112071168-112071168 on Build GRCh38

View Full Product Details