Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104861160-104861427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC41A2
Entrez Gene ID
84102
Gene Name
solute carrier family 41 member 2
Gene Aliases
SLC41A1-L1
UniGene
Hs.577463
Gene Chromosome Location
Chr.12: 104802553-104958744 on Build GRCh38
Transcripts
NM_032148.3 XM_005269176.1 XM_005269178.1 XM_005269179.1 XM_006719630.2 XM_011538807.2 XM_011538808.1 XM_011538809.1 XM_011538810.2 XM_011538811.2 XM_011538813.2 XM_017020013.1 XM_017020014.1 XM_017020016.1 XM_017020017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3376087 c.1255+3A>G p.? Chr.12: 104861288-104861288 on Build GRCh38
7390570 c.1184G>T p.G395V Chr.12: 104861362-104861362 on Build GRCh38
1746697 c.1182G>T p.G394G Chr.12: 104861364-104861364 on Build GRCh38
3772233 c.1006+7A>G p.? Chr.12: 104861284-104861284 on Build GRCh38
4727377 c.992C>T p.T331M Chr.12: 104861305-104861305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796612796 Chr.12: 104861179-104861179 on Build GRCh38
rs961295231 Chr.12: 104861266-104861266 on Build GRCh38
rs776892712 Chr.12: 104861368-104861368 on Build GRCh38
rs760809656 Chr.12: 104861361-104861361 on Build GRCh38
rs776188800 Chr.12: 104861372-104861372 on Build GRCh38
rs971276400 Chr.12: 104861184-104861184 on Build GRCh38
rs775635488 Chr.12: 104861305-104861305 on Build GRCh38
rs1028156834 Chr.12: 104861214-104861214 on Build GRCh38
rs927643162 Chr.12: 104861278-104861278 on Build GRCh38
rs749197572 Chr.12: 104861268-104861268 on Build GRCh38
rs762310756 Chr.12: 104861369-104861369 on Build GRCh38
rs991009650 Chr.12: 104861313-104861313 on Build GRCh38
rs374531137 Chr.12: 104861256-104861256 on Build GRCh38
rs953915026 Chr.12: 104861243-104861243 on Build GRCh38
rs111463634 Chr.12: 104861248-104861248 on Build GRCh38
rs371159491 Chr.12: 104861391-104861391 on Build GRCh38
rs772197079 Chr.12: 104861304-104861304 on Build GRCh38
rs758973052 Chr.12: 104861388-104861388 on Build GRCh38
rs754594779 Chr.12: 104861242-104861242 on Build GRCh38
rs764526539 Chr.12: 104861315-104861315 on Build GRCh38
rs1053565006 Chr.12: 104861275-104861275 on Build GRCh38
rs183969230 Chr.12: 104861239-104861239 on Build GRCh38
rs764318040 Chr.12: 104861364-104861364 on Build GRCh38
rs192915546 Chr.12: 104861384-104861384 on Build GRCh38
rs751955964 Chr.12: 104861325-104861325 on Build GRCh38
rs368725116 Chr.12: 104861377-104861377 on Build GRCh38
rs751029603 Chr.12: 104861381-104861381 on Build GRCh38
rs769752338 Chr.12: 104861249-104861249 on Build GRCh38

View Full Product Details